OPGAL EyeCGas CO Optical Gas Imaging Camera for Carbon Monoxide
Thermo Scientific™

OPGAL EyeCGas CO Optical Gas Imaging Camera for Carbon Monoxide

Detect dangerous CO leaks instantly with EyeCGas CO, a rugged, intrinsically safe OGI camera built for safety, speed, and reliability in industrial settings.
Have Questions?
Catalog NumberType
ECG30C-30Q-COMultigas Detector
Catalog number ECG30C-30Q-CO
Price (CLP)
-
Type:
Multigas Detector
The OPGAL EyeCGas™ CO Optical Gas Imaging Camera provides an intrinsically safe, rugged, and reliable solution for detecting carbon monoxide leaks in hazardous industrial environments. Engineered to identify CO remotely from a safe distance, the EyeCGas CO camera enhances safety, minimizes downtime, and helps prevent harmful emissions. With multi-spectral OGI capability, thermographic imaging, built-in quantification tools, and seamless connectivity, the EyeCGas CO camera supports fast leak identification, verification, and reporting across demanding oil, gas, and manufacturing applications like in the steel industry.

The EyeCGas CO camera combines advanced spectral filtering, rugged design, and high sensitivity thermal imaging to deliver outstanding performance for carbon monoxide leak detection.

>Key Capabilities and Benefits

  • CO leak detection: Quickly detects carbon monoxide and carbon dioxide for safety, compliance, and emissions monitoring.
  • Thermographic imaging: Supports temperature measurement and enhanced visualization using multiple color palettes.
  • Multi spectral OGI: Replaceable spectral filters optimized for CO, enabling superior target-specific detection.
  • Intrinsically safe: Certified for IECEx Zone II, ANSI, and CSA Class I & Class II Division 2 hazardous locations.
  • Built-in quantification: Realtime and offline CO quantification supported via built-in tools, EyeCSite software, and compatible LDAR systems.
  • Connectivity: Built-in Wi-Fi, hotspot, GPS, USB data transfer, and Bluetooth 4.2.
  • Rugged & sealed: Sealed to IP65 and engineered for gas leak detection in extreme oil & gas conditions.
  • LDAR ready: Integrates with TVA2020, RMLD, and other LDAR devices.
  • Free firmware upgrades: Receives ongoing software improvements based on customer feedback.

 

Specifications
Accuracy±1°C (0 to100°C); ±2% (>100°C); ±2°C (-20 to 0°C)
Battery Life>4.5 hours continuous operation
Battery TypeRechargeable Li-ion, 7.4 V
Certifications/Compliance
CSA C22.2 No. 213-M1987, Non-Incentive Electrical Equipment for Use in Class I, Division 2, ANSI/ ISA-12.12.01 – Class I and II, Division 2, and Class III, ATEX. Intrinsically safe for Zone 2 ratings as: Ex II 3 GD; Ex ic nA nC IIC T6 Gc; Ex ic tc IIIC T85°C DC
ColorRainbow, Iron, ISO red, ISO green, Grey Scale, Vivid
Detector TypeDetector pitch: 30 µm
Dimensions (L x W x H)230 x 110 x 130 mm (9 x 4.3 x 5.1 in.)
Display Type3.5 in. LCD, 640 x 480 pixels
IP RatingIP65
IncludesCamera, 2 x batteries, charger, USB cable, neck strap, glare shield, carrying case, cleaning kit
Mounting OptionsUNC 1/4 in. -20
ResolutionIR resolution: 320 × 240 pixels
Temperature Range (English)-4°F to 662°F
Temperature Range (Metric)-20°C to 350°C
Voltage7.4 V
Warranty4 years (Detector & Cooler: 2 years; Batteries: 1 year)
Weight (English)5.9 lb
Weight (Metric)2.6 kg
Components MeasuredCarbon Monoxide, Carbon Dioxide
TypeMultigas Detector
Unit SizeEach