Eutech™ Ion 6+ meter
Thermo Scientific™

Eutech™ Ion 6+ meter

Quick and easy measurement with this ion-selective meter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
ECION602PLUSKpH Electrode
ECION601PLUSATC Electrode
Catalog number ECION602PLUSK
Price (KRW)
-
Includes:
pH Electrode

Quick and easy measurement with this ion-selective meter. Applications for the Eutech™ Ion 6+ meter include: ISE (Use with any ISE with BNC connection), water quality testing, environmental/agricultural testing, laboratories and schools, and in industrial settings.

Features

  • Direct Ion Concentration for use with most mono- or di-valent Ion Selective Electrodes (ISE)
  • mV measurement mode allows you to check performance and calibration of your ISE
  • Multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Non-Volatile Memory ensures data are retained even meter powers off
  • Hold Function momentarily freezes reading for easy viewing and recording
  • Auto-Power Off after 17 minutes saves battery power after non-use
  • Self-Diagnostic with message codes for easy troubleshooting
  • Large Custom LCD provides optimum viewing even at a distance
  • Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
  • Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
  • pH Buffer Options: 1.68, 4.01, 7.00, 10.01, 12.45 (USA), 1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
Specifications
Accuracy (ISE)± 1% full scale
Accuracy (Temperature)±0.5°C
Accuracy (mV)1 mV & ±2 mV (beyond ±200 mV)
Accuracy (pH)± 0.01
Battery Type4 AAA x 1.4V
Calibration Points (pH)Up to 5 points
DescriptionEutech Ion 6+ Ion/pH/mV Meter with Single Junction pH Electrode ECFC7252101B, ATC Electrode ECPHTEM01P, & pH Carrying Kit (Includes 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution & 1 Deionised (Rinse) water (60 ml Each), Rubber Armour/Stand
IncludespH Electrode
Resolution (Temperature)0.1°C
Resolution (mV)0.1 mV and ±0.2 mV (within ±199.9 mV)
Calibration Points (ISE)Up to 5
Measurement ModesIon-Selective Meter
Range (ISE)Ion Concentration: 0.01 to 0.99 ppm, 1.0 to 199.9 ppm, 200 to 1999 ppm
Range (mV)±500 mV
Range (pH)0 to 14
Resolution (ISE)0.01 ppm, 0.1 ppm, 1 ppm
Resolution (pH)0.01
Temperature (Metric) Range0°C to 100.0°C
Unit SizeEach