Eutech™ PC 450 Multi-parameter Meter Kit
Actual product may vary
Thermo Scientific™

Eutech™ PC 450 Multi-parameter Meter Kit

Measure multiple parameters in a cost-effective and convenient way by using only one Eutech™ multi-parameter meter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
ECPCWP45003Meter with Combi Electrode
ECPCWP45000Multiparameter Meter
Catalog number ECPCWP45003
Price (HKD)
-
Product Type:
Meter with Combi Electrode

Your all-in-one solution for multi-parameter research work in the lab. The multi-parameter Eutech PC 450 measures pH / mV / Conductivity / Total Dissolved Solids / Temperature. It accepts a single pH/Con probe or separate pH and conductivity probes.

Features

  • Switch between pH, mV, conductivity and TDS with a press of a button
  • Large, backlit custom display – Lets you view measurement with temperature
  • Grip-Clip™ – Quickly and easily secures separate pH and conductivity electrodes simultaneously to your sample container or beaker
  • Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). New manual option allows calibration with up to 5 custom buffer standards you
  • Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
  • Intuitive, user-friendly icons – Electrode status displays pH slope or cell constant
  • Auto-range conductivity from 0 to 199.9 mS in five ranges – With up to five calibration points
  • Selectable cell constant – 0.10, 1.00 or 10.0
  • Adjustable temperature coefficient – From 0.00 to 10.00 % per ºC
  • Conductivity-to-TDS factor – Adjustable from 0.40 to 1.00
  • Selectable ºC/ºF – Choose units to best fit your application
  • Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
  • Store up to 500 data sets; easy USB or RS232 output with optional data cables
  • Built-in real-time clock stamps stored data and calibration data with time and date – Meets standards for GLP (Good Laboratory Practices)
  • Built-in stand with wall-mount – Allows for easy benchtop measurement and storage
Specifications
AC AdapterOptional universal power adapter
Accuracy (Conductivity)± 1% full scale
Accuracy (TDS)± 1% full scale
Accuracy (mV)±0.2 mV +2 LSD or +0.5 % of reading; whichever is greater
Accuracy (pH)±0.01
Alarm OutputCalibration due alarm
Calibration Points (Conductivity)Auto/manual; one point per range
Calibration Points (pH)Up to 6 points: USA, NIST, DIN or custom
CompensationTemperature: Automatic or manual from -10 to 110 °C
DescriptionPC 450 Meter & Combi Electrode: Meter, combi pH/Con/ATC electrode, two AA batteries
Dimensions (L x W x H)Meter: 7 7/8in x 3 1/8in x 2 1/4in (20 x 8.3 x 5.7 cm)
MemoryStores up to 500 data sets
OutputsUSB or RS232
Power SupplyTwo AA batteries (included), >100 hours (without backlight); AC adapter (optional)
Product TypeMeter with Combi Electrode
Resolution (Conductivity)0.01 μS to 0.1 mS
Resolution (TDS)0.01 ppm to 0.1 ppt
Resolution (mV)0.1 mV ±999.9 mV; 1 mV beyond
Temperature Accuracy±0.5°C, ±0.9°F
Temperature Range (English)14.0°F to 230.0°F
Temperature Resolution0.1 °C/°F
Weight (English)Meter: 1 lb
Weight (Metric)Meter: 0.5 kg
Display TypeLCD Graphic, Backlit
IP RatingIP67
Measurement ModespH, ORP, Ion, Conductivity, TDS, Salinity, Resistivity
Range (Conductivity)0 to 200 mS, 0 to 20 mS if using COMBI03BW probe
Range (TDS)0 to 200 ppt (TDS factor 1.0)
Range (mV)±2000 mV
Range (pH)-2 to 16
Resolution (pH)0.01
Temperature Range (Metric)-10.0°C to +110.0°C
Unit SizeEach