Eutech™ PD 450 Meter
Thermo Scientific™

Eutech™ PD 450 Meter

Collecting multi-parameter data is a breeze using Eutech™ PD 450 Meter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
ECPDWP45004Meter with Electrodes
ECPDWP45000Multiparameter Meter
Catalog number ECPDWP45004
Price (HKD)
-
Product Type:
Meter with Electrodes

The Eutech™ PD450 Meter is a multi-parameter meter that measures pH, mV, dissolved oxygen, and temperature. Affordable optical RDO technology reduces maintenance time and costs. The maintenance free optical probe with no electrolyte eliminates the hassle of replacing delicate membranes and salt build up from refillling solutions. You can also easily switch between pH, mV, % saturation and mg/L (ppm) dissolved oxygen – with the press of a button.

Features:

  • pH / mV / Dissolved Oxygen / Temperature
  • Durable waterproof and dustproof design with IP67 rating
  • Large, backlit graphic display provides measurement with temperature
  • Calibrate up to 5 points automatically with USA or NIST buffer groups (up to 6 points with DIN). New manual option allows calibration with up to 5 custom buffer standards
  • Auto buffer recognition – Automatically identifies the correct pH buffer for rapid calibration
  • Calibration due alarm – Set a visible reminder when it is time to recalibrate the meter
  • Selectable ºC or ºF – Choose units to best fit your application
  • Ready indicator, manual or automatic measurement hold feature and optional automatic shutoff function
  • Store up to 500 data sets – Easy USB or RS232 output with optional data cables
  • Built-in real-time clock stamps stored data and calibration data with time and date – Meets standards for GLP (Good Laboratory Practices)
Specifications
AC AdapterOptional
Accuracy (DO)±2 %; ±0.1 ppm or mg/L up to 8; ±0.2 ppm or mg/L above 8
Accuracy (mV)±0.2 mV +2 LSD or +0.5 % of reading; whichever is greater
Accuracy (pH)±0.01
Battery Life100+ hr. (without backlight)
Battery Type2 AA (included)
CalibrationmV: ±200 mV, DO: Zero and saturation, Temp: Offset in 0.1 increments, ±10 °C or &
Calibration Points (pH)Up to 6 points: USA, NIST, DIN or custom
CompensationPressure & Salinity Compensation: manual
DescriptionPD 450 Meter & Electrodes: Meter, pH/ATC electrode, RDO electrode with sensor cap, two AA batteries
Dimensions (L x W x H)Meter: 7 7/8in x 3 1/8in x 2 1/4in (20 x 8.3 x 5.7 cm), Boxed: 9 1/8in x 9 1/8in x 2 3/4in (23 x 23 x 7 cm)
IncludesTime and date stamp on calibration and stored data
MemoryStores up to 500 data sets
OutputsUSB or RS232
Product TypeMeter with Electrodes
Range (Polarographic DO)0 to 200 %; 0 to 20 ppm/mg/L
Resolution (Polarographic DO)0.10 %; 0.1 ppm/mg/L
Resolution (mV)0.1 mV ±999.9 mV; 1 mV beyond
Resolution (pH)0.01
Temperature Accuracy±0.5°C, ±0.9°F
Temperature CompensationAutomatic or manual from -10 to 110 °C
Temperature Range (English)14.0°F to 230.0°F
Temperature Resolution0.1 ºC/ºF
Weight (English)Meter: 1 lb , Boxed: 1.4 lb
Weight (Metric)Meter: 0.5 kg, Boxed: 0.7 kg
Display TypeBacklit Custom LCD
IP RatingIP67
Measurement ModespH, mV, Dissolved Oxygen, Temperature
Range (mV)±2000 mV
Range (pH)-2 to 16
Temperature Range (Metric)-10.0°C to +110.0°C
Unit SizeEach