Eutech™ pH 700 Meter
Actual product may vary
Eutech™ pH 700 Meter
Eutech™ pH 700 Meter
Thermo Scientific™

Eutech™ pH 700 Meter

Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
ECPH70042SpH electrode, ATC Probe
ECPH70040100/240 VAC Adapter
Catalog number ECPH70042S
Price (HKD)
-
Includes:
pH electrode, ATC Probe

Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools. It features a large, easy-to-read display, ready indicator that letsyou know when readings are stable and up to 5-point calibration with auto-buffer recognition. It also has a non-volatile memory holds up to 100 data points and comes witu an integral electrode holder.

Applications:

  • Educational: Useful for most laboratory, ecological studies and other applications.
  • Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing. Use in all types of food processing.

Buffer Sets

  • USA (1.68, 4.01, 7.00, 10.01, 12.45)
  • NIST (1.68, 4.01, 6.86, 9.18, 12.45)
  • Specifications
    Accuracy (Temperature)0.3°C/0.5°F
    Accuracy (mV)±0.2 (±199.9), ±2 (Beyond)
    Accuracy (pH)±0.01
    DescriptionEutech pH 700 Meter With pH Electrode (ECFC7252101B), ATC Probe (PH5TEMB01P), Integral Electrode Holder and 100/240 VAC Adapter
    IncludespH electrode, ATC Probe
    TypepH Meter
    Calibration PointsUp to 5
    Resolution (pH)0.01
    Temperature Range (Metric)0°C to 100°C
    Unit SizeEach