Heratherm CO<sub>2</sub> Chamber 750 L
Heratherm CO<sub>2</sub> Chamber 750 L
Heratherm CO<sub>2</sub> Chamber 750 L
Heratherm CO<sub>2</sub> Chamber 750 L
Heratherm CO<sub>2</sub> Chamber 750 L
Heratherm CO<sub>2</sub> Chamber 750 L
Heratherm CO<sub>2</sub> Chamber 750 L
Heratherm CO<sub>2</sub> Chamber 750 L
Heratherm CO<sub>2</sub> Chamber 750 L
Thermo Scientific™

Heratherm CO2 Chamber 750 L

Precisely control the essential parameters of your distinct cell growth or CO2 testing conditions — active humidity controls the incubation environment with accuracy.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDoor StyleCapacity (Metric)
ECS750Solid (outer), Glass (inner)750 L
ECG750Glass door750 L
ECS250Solid outer door with inner glass door247 L
ECG250Glass door247 L
Catalog number ECS750
Price (USD)
-
Door Style:
Solid (outer), Glass (inner)
Capacity (Metric):
750 L
Accurate control of three parameters - Temperature above and below ambient, relative humidity and CO2 - provide great environment in Cell growth and testing applications for reproducible results.

 

  • Uniform Temperature / CO2 / relative humidity throughout the chamber
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • CO2 control system ensures stable CO2 environment, range of 0 – 20 Vol-%.
  • Environmentally friendly water-efficient humidification system - with water tank or optional fixed water connection for in-house purified water
  • Intuitive touchscreen interface with easy programmability of parameter cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with passcode; restrict access with standard key door lock or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option.
Choose from a variety of country-specific sockets. (Ordered with separate order code)

 

Specifications
Alarm TypesAudible and Visible
Capacity (English)26.5 cu.ft.
Capacity (English) Shelf66 lbs. (stationary)
Capacity (Metric) Shelf30 kg (stationary)
Certifications/ComplianceUL, CE
Data OutputsUSB, Thermo Cloud via OPC UA
Dimensions (L x W x H)895 x 1080 x 1835 mm / 35.24 x 42.52 x 72.24 in.
Dimensions (LxWxH) Interior630 x 840 x 1450 mm / 24.8 x 33.07 x 57.09 in.
Display7 in. Touchscreen user interface
Door StyleSolid (outer), Glass (inner)
Electrical Requirements100/240 V, 50/60 Hz, 1400 W
For Use With (Application)Cell growth, testing
Humidity Range10 - 90% RH
Humidity Resolution0.1% RH
Humidity SourceRemovable water tank
No. of Casters4
Plug TypeNema 5-20P
Ports2 access ports
ProgrammabilityTemperature, relative humidity, CO2 ramping
Shelf Area (English)4.9 ft.2
Shelf Area (Metric)0.46 m2
Shelf MaterialStainless Steel (304/1.4301)
Shipping Weight (English)859 lbs.
Shipping Weight (Metric)390 kg
Temperature ControlMicroprocessor - programmable
TypeGrowth Chamber
Warranty24 months
Water Consumption<0.8 L per day - Value depends on settings, usage (e.g. number of door openings), and environmental conditions
Water Resistivity0.05 - 1 MΩ
Wattage1400 W
Width (English) Shelf31.3 in.
Width (Metric) Shelf795 mm
Capacity (Metric)750 L
Chamber MaterialStainless Steel (304/1.4301)
MaterialPowder-coated Galvanized Steel
No. of Shelves3
Relative Humidity10 to 90%
Voltage100/115/220/230 V
Unit SizeEach