Connect Edge Adapter Kit for InstrumentConnect Remote Monitoring with OPC UA™
Thermo Scientific™
Connect Edge Adapter Kit for InstrumentConnect Remote Monitoring with OPC UA™
Connect Edge Wi-Fi/Ethernet device adapters are for use with InstrumentConnect 24/7 remote monitoring system. Device adapters plug into Thermo Fisher laboratory equipment to extract and transmit telemetry to the Connect Edge gateway via Wi-Fi or Ethernet. Configuration needed varies by equipment type.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)Includes
EDGEKTADPT05Wireless Monitoring KitConnect Edge SensorsWi-Fi Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3 m Cable, Power Supply and Country Converters. Convert to Ethernet with EDGEKTADPT99 (no PoE)
EDGEKTADPT01Remote Monitoring Device AdapterConnect Edge Hardware, CO2/ULT Equipment and DevicesConnect Edge Wi-Fi Device Adapter Kit with RS485, (optional Ethernet when combined with EDGEKTADAPT99)
EDGEKTADPT02Remote Monitoring Device AdapterConnect Edge Hardware, Refrigerators/Freezers and DevicesWi-FI (optional Ethernet when combined with EDGEKTADAPT99)
EDGEKTADPT01BWireless Monitoring KitCO2, BSC and ULT, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with RS485 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m (depending on kit), power supply mounting bracket.
EDGEKTADPT03Remote Monitoring Device AdapterConnect Edge, CO2 Incubators and DevicesWi-FI (optional Ethernet when combined with EDGEKTADAPT99)
EDGEKTADPT02BWireless Monitoring KitRefrigerator/Freezer, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with TTL connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable and 2-3 m (depending on kit), power supply, mounting bracket.
EDGEKTADPT04Remote Monitoring Device AdapterConnect Edge, CO2 Incubators and DevicesWi-FI (optional Ethernet when combined with EDGEKTADAPT99)
EDGEKTADPT03BWireless Monitoring KitRefrigerator/Freezer, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with RS232 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable and 2-3 m (depending on kit), power supply, mounting bracket.
EDGEKTADPT99Remote Monitoring Device AdapterConnect Edge, Laboratory Equipment and DevicesOptional PoE Converter for Wi-Fi/Ethernet Adapters
EDGEKTADPT04BWireless Monitoring KitIncubators, Environmental Chambers, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with USB connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m (depending on kit), power supply, mounting bracket.
EDGEKTADPT51Remote Monitoring Device AdapterConnect Edge, ULT/CO2 Incubators and DevicesEthernet-Only Connect Edge Device Adapter Kit with RS485. Only for use in: Brazil, UAE, Bahrain, Oman
EDGEKTADPT51BWireless Monitoring KitCO2, BSC and ULT, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with RS485 connection, power cable 2-3 m (depending on kit), power supply, mounting bracket.
EDGEKTADPT52Remote Monitoring Device AdapterConnect Edge, Refrigerators/Freezers and DevicesEthernet-Only Connect Edge Device Adapter Kit with TTL. Only for use in: Brazil, UAE, Bahrain, Oman
EDGEKTADPT52BWireless Monitoring KitRefrigerator/Freezer, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with TTL connection, power cable 2-3 m (depending on kit), power supply, mounting bracket.
EDGEKTADPT53Remote Monitoring Device AdapterConnect Edge, CO2 Incubators and DevicesEthernet-Only Connect Edge Device Adapter Kit with RS232. Only for use in: Brazil, UAE, Bahrain, Oman
EDGEKTADPT53BWireless Monitoring KitRefrigerator/Freezer, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with RS232 connection, power cable 2-3 m (depending on kit), power supply, mounting bracket.
EDGEKTADPT54Remote Monitoring Device AdapterConnect Edge, CO2 Incubators and DevicesEthernet-Only Connect Edge Device Adapter Kit with USB. Only for use in: Brazil, UAE, Bahrain, Oman
EDGEKTADPT54BWireless Monitoring KitIncubators, Environmental Chambers, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with USB connection, power cable and 2-3 m (depending on kit), power supply, mounting bracket.
EDGEKTADPT55Wireless Monitoring KitConnect Edge SensorsWi-Fi Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3 m power cable, powers supply and country converters.
EDGEKTADPT01PBWireless Monitoring KitCO2, BSC and ULT, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with RS485 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
EDGEKTADPT02PBWireless Monitoring KitRefrigerator/Freezer, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with TTL connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
EDGEKTADPT03PBWireless Monitoring KitRefrigerator/Freezer, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with RS232 connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
EDGEKTADPT04PBWireless Monitoring KitIncubators, Environmental Chambers, See thermofisher.com/edgecompatible for model detailsConnect Edge Wi-Fi Device Adapter Kit with USB connection, (Purchase EDGEKTADAPT99 for Ethernet/PoE), power cable 2-3 m, mounting bracket.
EDGEKTADPT51PBWireless Monitoring KitCO2, BSC and ULT, See thermofisher.com/edgecompatible for model detailsConnect Edge Ethernet Device Adapter Kit with RS485 connection, power cable 2-3 m, mounting bracket.
EDGEKTADPT52PBWireless Monitoring KitRefrigerator/Freezer, See thermofisher.com/edgecompatible for model detailsConnect Edge Ethernet Device Adapter Kit with TTL connection, power cable 2-3 m (depending on kit), mounting bracket, no power supply
EDGEKTADPT53PBWireless Monitoring KitRefrigerator/Freezer, See thermofisher.com/edgecompatible for model detailsConnect Edge Ethernet Device Adapter Kit with RS232 connection, power cable 2-3 m, mounting bracket.
EDGEKTADPT54PBWireless Monitoring KitIncubators, Environmental Chambers, See thermofisher.com/edgecompatible for model detailsConnect Edge Ethernet Device Adapter Kit with USB connection, power cable 2-3 m, mounting bracket.
EDGEKTADPT05PRemote Monitoring Sensor AdapterConnect Edge SensorsWi-Fi Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3M power cable. Convert to Ethernet with EDGEKTADPT99 (no PoE)
EDGEKTADPT55PRemote Monitoring Sensor AdapterConnect Edge SensorsEthernet Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3M Cable
Catalog number EDGEKTADPT05
Price (KRW)
-
Type:
Wireless Monitoring Kit
For Use With (Equipment):
Connect Edge Sensors
Includes:
Wi-Fi Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3 m Cable, Power Supply and Country Converters. Convert to Ethernet with EDGEKTADPT99 (no PoE)
Connect Edge Wi-Fi/Ethernet device adapters are part of the InstrumentConnect 24/7 remote monitoring system. Device adapters include RS485, TTL, RS232 and USB configurations to plug into your Thermo Fisher laboratory equipment. Device adapters extract and transmit telemetry to the Connect Edge gateway via Wi-Fi or Ethernet.
Thermo Scientific™ InstrumentConnect is professional-grade remote monitoring system for monitoring laboratory equipment and other devices to help protect samples and increase asset management efficiencies. The InstrumentConnect system utilizes Connect Edge adapters and gateways to send data to the InstrumentConnect cloud or to your own premise-based system (LIMS, BMS, SCADA) using OPC-UA™ protocol. Connect Edge hardware is compatible with 700+ Thermo Fisher Scientific laboratory equipment makes and models.
Thermo Fisher Scientific Equipment Telemetry
  • For cold storage equipment, InstrumentConnect collects ambient and cabinet temperature data recorded by resistance temperature detectors (RTDs) built into the unit; the remote monitoring system also reports alarms and events, as well as other system-based diagnostic data points to the InstrumentConnect application.
  • For CO2 incubators and environmental chambers, InstrumentConnect collects chamber temperatures, gas levels, alarms and alerts received via the incubator and environmental chamber data output connection.
System Hardware
  • Device Adapter-Palm-size adapters collect and transmit data from lab equipment/devices to gateways using Ethernet (WAN) connectivity or Wi-Fi.
  • Connect Edge Gateway-Receives data from device adapters to send to InstrumentConnect (cloud) or customer-premise system/cloud via OPC-UA using Ethernet (WAN) connectivity or Wi-Fi.
System Software
Choose from these available InstrumentConnect packages.
InstrumentConnect Monitor (Cloud):
  • Monitor onboard telemetry data and events**
  • Custom alarm settings
  • View live device status 24/7 via Mobile and Cloud app
  • Peace of mind with industry-leading security protocols
  • 25GB included data storage for documents
  • Unlimited device telemetry storage
  • Notifications via mobile application, e-mail and desktop application
InstrumentConnect Integrate (OPC-UA to Cloud or Prem):
  • Utilize the Connect Edge system to collect and transmit data to your on-premise system or cloud using Open Platform Communications Unified Platform. The Connect Edge gateway can be enabled as an OPC server for 10 or more devices.
Network
You can connect 10 device adapters to each Connect Edge gateway. Gateways can accommodate greater than 10 devices, however this is determined by reviewing CPU usage after you have reached this initial quantity after initial set up. Deciding which topology is applicable to your setting will determine the number of gateways you will need.
Connect Edge system has 3 main topologies
  • Direct Wi-Fi-Device adapters are configured to transmit data directly to your Connect Edge gateway (10 max per gateway). Your device adapters must be in proximity (50-150 feet) to the gateway.
  • Single/Dual Network-Device adapters are configured to connect to your local network via Ethernet or Wi-Fi and transmit data to the Connect Edge gateway, also on the same network. This topology is ideal if devices to be monitored are not within proximity to each other or a gateway.
Building your system
  • Determine type (Ethernet-only/WiFi+Ethernet) and quantity of gateway(s) needed based on network topology preference. The Configuration Planning Guide can be used for reference, if needed.
  • Select compatible adapter for your lab equipment
  • Purchase an InstrumentConnect Monitor or Integrate subscription for each device
  • Determine whether self-install or professional-install is desired (where available)
System hardware and software is available for purchase in packages or a-la-carte. A built-in gateway discount is included with all packages. Additional savings with 3 and 5-year packages.
Installation
Customer (self) installation
Installation services sold separately, if desired (US, Europe, Great Britain, inquire with specialist for other regional availability).
Availability
InstrumentConnect is available in the following countries and configurations. For the most updated list, please ask your specialist.
Wi-Fi or Ethernet gateway/adapter: Europe, UK, North America, Australia, New Zealand, Puerto Rico
Ethernet-only gateway/Wi-Fi or Ethernet adapter: Hong Kong
Ethernet-only gateway and adapter: Brazil, Bahrain, Oman, UAE
Specifications
Battery TypeNone
CalibrationNA
Certifications/CompliancePower Adapter: CE, UKCA, FCC, RCM, CCC, UL, BIS, WEEE and ROHS
Data MemoryNone
For Use With (Equipment)Connect Edge Sensors
IP RatingIP66/68
IncludesWi-Fi Sensor Adapter for Independent Sensors. Ports for up to 6 analog sensors and 1 digital sensor. Includes 3 m Cable, Power Supply and Country Converters. Convert to Ethernet with EDGEKTADPT99 (no PoE)
ModelIncludes Power Supply
Quantity1
Warranty1 Year Warranty
Alarm TypesNone
Display TypeNo Display
Software TypeInstrumentConnect
TypeWireless Monitoring Kit
Unit SizeEach