InstrumentConnect Monitor Subscription for 24/7 Laboratory Remote Monitoring (No gateway))
Thermo Scientific™
InstrumentConnect Monitor Subscription for 24/7 Laboratory Remote Monitoring (No gateway))
InstrumentConnect Monitor Subscriptions provide access to the InstrumentConnect desktop or mobile application for 24/7 remote monitoring of laboratory equipment. Requires Connect Edge gateway(s) and adapter(s).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
EDGM3D5Y3 Monitor Subscription, 5 years
EDGM1D1Y1 Monitor Subscription, 1 year
EDGM1D2Y1 Monitor Subscription, 2 years
EDGM1D3Y1 Monitor Subscription, 3 years
EDGM1D5Y1 Monitor Subscription, 5 years
EDGM3D2Y3 Monitor Subscription, 2 years
EDGM5D2Y5 Monitor Subscription, 2 years
EDGM10D2Y10 Monitor Subscription, 2 years
EDGM15D2Y15 Monitor Subscription, 2 years
EDGM20D2Y20 Monitor Subscription, 2 years
EDGM50D2Y50 Monitor Subscription, 2 year
EDGM3D3Y3 Monitor Subscription, 3 years
EDGM5D3Y5 Monitor Subscription, 3 years
EDGM10D3Y10 Monitor Subscription, 3 years
EDGM15D3Y15 Monitor Subscription, 3 years
EDGM20D3Y20 Monitor Subscription, 3 years
EDGM50D3Y50 Monitor Subscription, 3 year
EDGM5D5Y5 Monitor Subscription, 5 years
EDGM10D5Y10 Monitor Subscription, 5 Years
EDGM15D5Y15 Monitor Subscription, 5 years
EDGM20D5Y20 Monitor Subscription, 5 years
EDGM50D5Y50 Monitor Subscription, 5 year
Catalog number EDGM3D5Y
Price (CLP)
-
Includes:
3 Monitor Subscription, 5 years
InstrumentConnect is an agnostic, professional grade 24/7 remote monitoring system for laboratory equipment and devices designed to help protect and manage vital products and samples. This system features customizable parameters, live alarms, notifications and cost-effective Connect Edge hardware that can collect data from 10 or more instruments. InstrumentConnect Monitor Subscriptions provide access to remote cloud monitoring through desktop or mobile application. Subscriptions may be purchased a-la-carte or as part of a gateway + subscription package for greater savings. One subscription per device.
Thermo Scientific™ InstrumentConnect is professional-grade remote monitoring system for monitoring laboratory equipment and other devices to help protect samples and increase asset management efficiencies. The InstrumentConnect system utilizes Connect Edge adapters and gateways to send data to the InstrumentConnect cloud or to your own premise-based system (LIMS, BMS, SCADA) using OPC-UA™ protocol. Connect Edge hardware is compatible with 700+ Thermo Fisher Scientific laboratory equipment makes and models.
Thermo Fisher Scientific Equipment Telemetry
  • For cold storage equipment, InstrumentConnect collects ambient and cabinet temperature data recorded by resistance temperature detectors (RTDs) built into the unit; the remote monitoring system also reports alarms and events, as well as other system-based diagnostic data points to the InstrumentConnect application.
  • For CO2 incubators and environmental chambers, InstrumentConnect collects chamber temperatures, gas levels, alarms and alerts received via the incubator and environmental chamber data output connection.
System Hardware
  • Device Adapter-Palm-size adapters collect and transmit data from lab equipment/devices to gateways using Ethernet (WAN) connectivity or Wi-Fi.
  • Connect Edge Gateway-Receives data from device adapters to send to InstrumentConnect (cloud) or customer-premise system/cloud via OPC-UA using Ethernet (WAN) connectivity or Wi-Fi.
System Software
Choose from these available InstrumentConnect packages.
InstrumentConnect Monitor (Cloud):
  • Monitor onboard telemetry data and events**
  • Custom alarm settings
  • View live device status 24/7 via Mobile and Cloud app
  • Peace of mind with industry-leading security protocols
  • 25GB included data storage for documents
  • Unlimited device telemetry storage
  • Notifications via mobile application, e-mail and desktop application
InstrumentConnect Integrate (OPC-UA to Cloud or Prem):
  • Utilize the Connect Edge system to collect and transmit data to your on-premise system or cloud using Open Platform Communications Unified Platform. The Connect Edge gateway can be enabled as an OPC server for 10 or more devices.
Network
You can connect 10 device adapters to each Connect Edge gateway. Gateways can accommodate greater than 10 devices, however this is determined by reviewing CPU usage after you have reached this initial quantity after initial set up. Deciding which topology is applicable to your setting will determine the number of gateways you will need.
Connect Edge system has 3 main topologies
  • Direct Wi-Fi-Device adapters are configured to transmit data directly to your Connect Edge gateway (10 max per gateway). Your device adapters must be in proximity (50-150 feet) to the gateway.
  • Single/Dual Network-Device adapters are configured to connect to your local network via Ethernet or Wi-Fi and transmit data to the Connect Edge gateway, also on the same network. This topology is ideal if devices to be monitored are not within proximity to each other or a gateway.
Building your system
  • Determine type (Ethernet-only/WiFi+Ethernet) and quantity of gateway(s) needed based on network topology preference. The Configuration Planning Guide can be used for reference, if needed.
  • Select compatible adapter for your lab equipment
  • Purchase an InstrumentConnect Monitor or Integrate subscription for each device
  • Determine whether self-install or professional-install is desired (where available)
System hardware and software is available for purchase in packages or a-la-carte. A built-in gateway discount is included with all packages. Additional savings with 3 and 5-year packages.
Installation
Customer (self) installation
Installation services sold separately, if desired (US, Europe, Great Britain, inquire with specialist for other regional availability).
Availability
InstrumentConnect is available in the following countries and configurations. For the most updated list, please ask your specialist.
Wi-Fi or Ethernet gateway/adapter: Europe, UK, North America, Australia, New Zealand, Puerto Rico
Ethernet-only gateway/Wi-Fi or Ethernet adapter: Hong Kong
Ethernet-only gateway and adapter: Brazil, Bahrain, Oman, UAE
Specifications
DescriptionInstrumentConnect Monitor Subscription
For Use With (Application)InstrumentConnect Remote Monitoring System, Laboratory Equipment and Devices
For Use With (Equipment)Connect Edge, Laboratory Equipment and Devices
IP RatingNone
Includes3 Monitor Subscription, 5 years
Quantity1 Ea.
Alarm TypesNone
TypeSoftware and Subscription Service
Unit SizeEach