Alkaline Phosphatase (ALP) Activity Kit
Invitrogen™

Alkaline Phosphatase (ALP) Activity Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
製品番号(カタログ番号)数量
EEA00296 tests
製品番号(カタログ番号) EEA002
価格(JPY)
70,000
キャンペーン価格
Ends: 25-Sep-2026
116,700
割引額 46,700 (40%)
Each
数量:
96 tests

The Alkaline Phosphatase (ALP) Activity Kit is a highly reliable and sensitive assay designed for the quantitative measurement of ALP activity in various sample types. ALP is a group of cytomembrane-related enzymes with hydrolysis and transfer activity, acting on a variety of phosphate substrates. ALP is a homologous dimerase where each catalytic site contains three metal ions.

The assay is based on the ability of alkaline phosphatase to decompose benzene disodium phosphate to produce free phenol and phosphoric acid. Phenol reacts with 4-aminopyrine in alkaline solution and oxidizes with potassium ferricyanide to form a red quinone derivative. The enzyme activity can be calculated indirectly by measuring the OD value.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

仕様
アッセイ範囲0.13–50 King Units/100 mL
アッセイ感度0.13 King Units/100 mL
使用対象 (装置)マイクロプレートリーダー
インキュベーション時間15 min.
アッセイ間のCV8.5
アッセイ内CV5.1
数量96 tests
サンプル量5 μL
出荷条件Dry Ice, 2–8°C
指名標的キットAlkaline Phosphatase (ALP)
総アッセイ時間30 min.
検出法比色法
サンプルタイプPlasma, Serum, Tissue Homogenates, Whole Cells
Unit SizeEach
組成および保存条件
Store at 2–8°C.