Cell Total Iron Colorimetric Assay Kit
Invitrogen™

Cell Total Iron Colorimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA00996 tests
Catalog number EEA009
Price (USD)
688.65
Online exclusive
710.00
Save 21.35
Each
Quantity:
96 tests
Price (USD)
688.65
Online exclusive
710.00
Save 21.35
Each

The Cell Total Iron Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of total iron content in cells. Iron plays an important role in various physiological functions. The ferrous ion is a key element in heme and hemoglobin and is present in many biochemical reactions.

In this assay iron ions in samples can be reduced into ferrous ions (Fe2+) when subjected to reductants. The latter then bind to probes in solution to form complexes that have a maximum absorption peak at 593 nm. The concentration of iron can be indirectly calculated by measuring the OD value.

Key features
• Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
• Broad range of kits—across key analytes and targets within the most important metabolic pathways
• Consistent—strict quality control criteria to ensure reproducible and reliable results

Specifications
Assay Range0.4–50 μmol/L
Assay Sensitivity0.4 μmol/L
For Use With (Equipment)Microplate Reader
Incubation Time40 min.
Interassay CV1.5
Intraassay CV1.3
Quantity96 tests
Sample Volume80 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedCell Total Iron
Total Assay Time80 min.
Detection MethodColorimetric
Sample TypeWhole Cells
Unit SizeEach
Contents & Storage
Store at 2–8°C.