High-density Lipoprotein Cholesterol (HDL-C) Colorimetric Assay Kit
Invitrogen™
High-density Lipoprotein Cholesterol (HDL-C) Colorimetric Assay Kit
These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Número de catálogoCantidad
EEA01296 pruebas
Número de catálogo EEA012
Precio (MXN)
15,765.19
Each
Cantidad:
96 pruebas
Precio (MXN)
15,765.19
Each
The High-density Lipoprotein Cholesterol (HDL-C) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of HDL-C in serum, plasma, culture supernatant, tissue, and cells. HDL-C is an anti-atherosclerotic lipoprotein that can transport cholesterol from extrahepatic tissues to the liver for metabolism and excretion of bile from the body. Its plasma level is negatively correlated with the risk of cardiovascular disease.
The assay is based on the reaction of HDL-C with enzyme reagents containing cholesterol esterase and cholesterol oxidase to produce hydrogen peroxide. The hydrogen peroxide is catalyzed by an oxidase in the presence of 4-aminoantipyrine (4-AA) and phenol (T-OOS) to form a red quinone compound. The colored substance has a maximum absorption peak at 546 nm. The HDL-C content in the sample can be calculated by measuring the OD value at 546 nm.
Key features
  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Especificaciones
Intervalo del ensayo0.065–3.8 mmol/L
Sensibilidad del ensayo0.065 mmol/L
Para utilizar con (equipo)Lector de microplacas
Tiempo de incubación10 min.
CV interensayo5
CV intraensayo3
Cantidad96 pruebas
Volumen de muestra2.5 μL
Condiciones de envíoDry Ice, 2–8°C
Nombre del kit de destinoHigh-density Lipoprotein Cholesterol (HDL-C)
Tiempo total del ensayo20 min.
Método de detecciónColorimétrico
Tipo de muestraCell Culture Supernatant, Plasma, Serum, Tissue Homogenates, Whole Cells
Unit SizeEach
Contenido y almacenamiento
Store at 2–8°C.