Non-esterified Free Fatty Acids (NEFA/FFA) Colorimetric Assay Kit
Invitrogen™

Non-esterified Free Fatty Acids (NEFA/FFA) Colorimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA01796 tests
Catalog number EEA017
Price (MXN)
-
Quantity:
96 tests

The Non-esterified Free Fatty Acids (NEFA/FFA) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of NEFA in animal tissue samples. NEFAs are derived from adiopose tissue, where hormone-sensitive lipase decomposes triglycerides to product fatty acids. These acids circulate freely in the body and, when combined with plasma albumin, can be used as an energy sourced for muscles, organs, and the brain. They are also a substrate of fat synthesis, where the concentration of NEFAs is related to metabolism and endocrine function.

In this assay NEFA react with nantokite to form copper soap, which has a specific absorption peak at 715 nm. The content of NEFA can be calculated indirectly by measuring the OD value.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range0.15–1.5 mmol/L
Assay Sensitivity0.15 mmol/L
For Use With (Equipment)Microplate Reader
Incubation Time3 min.
Interassay CV5.1
Intraassay CV3.3
Quantity96 tests
Sample Volume1000 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedNon-esterified Free Fatty Acids (NEFA/FFA)
Total Assay Time30 min.
Detection MethodColorimetric
Sample TypeTissue Homogenates
Unit SizeEach
Contents & Storage
Store at 2–8°C.