Reduced Glutathione (GSH) Colorimetric Assay Kit
Invitrogen™

Reduced Glutathione (GSH) Colorimetric Assay Kit

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
Catalog NumberQuantity
EEA02096 tests
Catalog number EEA020
Price (USD)
710.00
Each
Quantity:
96 tests
Price (USD)
710.00
Each

The Reduced Glutathione (GSH) Colorimetric Assay Kit is a highly reliable and sensitive assay designed for the quantitative measurement of GSH in serum, plasma, cells, cell culture supernatant, and tissue samples. GSH is a critical antioxidant molecule that plays a crucial role in maintaining cellular health and protecting against oxidative stress. GSH plays an important role in the research of prevention, recovery, and treatment of atherosclerosis, coronary heart disease, cancer, Alzheimer's, and other diseases.

In this assay, reduced GSH reacts with dinitrobenzoic acid to form a yellow complex in solution. GSH content can be calculated indirectly by measuring the OD value at 450 nm.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
Specifications
Assay Range2–100 μmol/L
Assay Sensitivity2 μmol/L
For Use With (Equipment)Microplate Reader
Incubation Time5 min.
Interassay CV3.2
Intraassay CV1.9
Quantity96 tests
Sample Volume100 μL
Shipping ConditionDry Ice, 2–8°C
Target Kit NamedReduced Glutathione (GSH)
Total Assay Time25 min.
Detection MethodColorimetric
Sample TypeCell Culture Supernatant, Plasma, Serum, Tissue Homogenates, Whole Cells
Unit SizeEach
Contents & Storage
Store at 2–8°C.