Total Cholesterol (TC) Colorimetric Assay Kit (single reagent, COD-PAP method)
Invitrogen™

Total Cholesterol (TC) Colorimetric Assay Kit (single reagent, COD-PAP method)

These complete, ready-to-use kits include clear 96-well plate(s), other assay specific reagents, buffers, and a lot-specific technical data sheet.
Have Questions?
카탈로그 번호수량
EEA02696 tests
카탈로그 번호 EEA026
제품 가격(KRW)
936,000
온라인 행사
Ends: 30-Sep-2026
1,069,000
할인액 133,000 (12%)
Each
수량:
96 tests
제품 가격(KRW)
936,000
온라인 행사
Ends: 30-Sep-2026
1,069,000
할인액 133,000 (12%)
Each

The Total Cholesterol (TC) Colorimetric Assay Kit (single reagent, COD-PAP method) is a highly reliable and sensitive assay designed for the quantitative measurement of total cholesterol in serum, plasma, and animal tissue. Cholesterol is a type of sterol and lipid found in cell membranes. It is an essential component of cell structure and function, but high levels of TC in the blood have been linked to an increased risk of cardiovascular disease. The measurement of TC levels (which includes free cholesterol and cholesterol esters) is an important diagnostic tool in clinical and research settings.

The assay utilizes the COD-PAP method, which involves the hydrolysis of cholesterol esters in the sample to produce free cholesterol and free fatty acids. Free cholesterol is then oxidized by cholesterol oxidase to produce 4-cholestenone and hydrogen peroxide. In the presence of 4-aminoamylpyridine and phenol, hydrogen peroxide catalyzes peroxidase to form red quinone compounds. The color depth of the generated quinone is directly proportional to the cholesterol content, which can be measured spectrophotometrically at 510 nm.

Key features

  • Simple and fast protocol – user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits – across key analytes and targets within the most important metabolic pathways
  • Consistent – strict quality control criteria to ensure reproducible and reliable results
사양
어세이 범위0.29 to 25.85 mmol/L
어세이 민감성0.29 mmol/L
용도(장비)Microplate Reader
인큐베이션 시간10 min.
분석 간 CV8.3
분석 내 CV3.1
수량96 tests
샘플 부피2.5 μL
배송 조건Dry Ice, 2–8°C
타겟 키트 이름 지정Total Cholesterol (TC)
총 어세이 시간20 min.
검출 방법Colorimetric
샘플 종류Plasma, Serum, Tissue Homogenates
Unit SizeEach
구성 및 보관
Store at 2°C to 8°C.

자주 묻는 질문(FAQ)

Can I use the Total Cholesterol (TC) Colorimetric Assay Kit (single reagent, COD-PAP method) (Cat. No. EEA026) with cell samples?

We do not recommend using the Total Cholesterol (TC) Colorimetric Assay Kit with cell samples because most untreated cell samples have a low TC that does not meet the sensitivity of the kit.

Find additional tips, troubleshooting help, and resources within our Protein Biology Support Centers .