Tartrate Resistant Acid Phosphatase (TRAP) Activity Assay Kit
Invitrogen™

Tartrate Resistant Acid Phosphatase (TRAP) Activity Assay Kit

With this kit, a chromogenic agent is catalyzed by acid phosphatase to produce p-nitrophenol in acidic condition.
Have Questions?
Catalog NumberQuantity
EEA05596 Tests
Catalog number EEA055
Price (USD)
696.00
Each
Quantity:
96 Tests
Price (USD)
696.00
Each

With this kit, a chromogenic agent is catalyzed by acid phosphatase to produce p-nitrophenol in acidic condition. P-nitrophenol has a maximum absorption at 405 nm. The activity of tartrate-resistant acid phosphatase (TRAP) can be calculated by measuring the amount of produced p-nitrophenol. The activity of acid phosphatase being detected in the presence of tartric acid is regarded as TRAP activity. This kit is useful for researchers studying bone metabolism and osteoclast activity.

Key features

  • Simple and fast protocol—user-friendly, requiring minimal hands-on time and providing rapid results within a short incubation period
  • Broad range of kits across key analytes and targets within the most important metabolic pathways
  • Consistent—strict quality control criteria to ensure reproducible and reliable results
For Research Use Only.
Specifications
Assay Range0.911 to 100 U/L
Assay Sensitivity0.911 U/L
For Use With (Equipment)Microplate Reader
Incubation Time2 min.
Interassay CV2.4%
Intraassay CV2.0%
Product TypeKit
Quantity96 Tests
Sample Volume20 μL
Shipping ConditionDry Ice, 2°C to 8°C
Target Kit NamedTartrate Resistant Acid Phosphatase (TRAP)
Total Assay Time15 min.
Detection MethodColorimetric
Immunoassay Kit FormatColorimetric Detection Kit
Sample TypeSerum, Plasma, Urine, Animal Tissue
Target SpeciesUniversal
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.