What is the purpose of the E-Holder Platform?
The E-Holder Platform is designed to hold E-PAGE Gels during loading. You can use the E-Holder Platform when you need to load multiple gels while other gels are running on the E-Base device.
Note: The E-Holder Platform is not a power supply unit, cannot be connected to an electrical outlet, and cannot be used to run E-PAGE 48 or 96 Gels. To obtain the best results, run E-PAGE Gels on the Mother E-Base Device or Daughter E-Base Device within 15 minutes after loading on the E-Holder Platform.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.
Note: The E-Holder Platform is not a power supply unit, cannot be connected to an electrical outlet, and cannot be used to run E-PAGE 48 or 96 Gels. To obtain the best results, run E-PAGE Gels on the Mother E-Base Device or Daughter E-Base Device within 15 minutes after loading on the E-Holder Platform.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.