This product is on backorder. Please see alternate Cat. No. KHC0121.

IL-12 (total) ELISA Kit, Human

Catalog number: EH2IL12T

Thermo Scientific™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Assay IL-12 p40 + p70 (total human interleukin-12) levels in human serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with these hermo Scientific Human IL-12 (total) ELISA Kits.

Features of this Human IL-12 ELISA Kit:

Target: total (p70 and p40) natural and recombinant human interleukin-12 (IL-12)
Format: colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: absorbance at 450nm minus absorbance at 550nm
Assay Range: 25.6 to 1000 pg/mL
Sensitivity: < 5 pg/mL
Precision: intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 3 hours
Sample Size: 50 µL
Specificity: no significant cross-reaction with human GM-CSF, IFNα, IFNγ, TNFα, TNFβ, IL-1α, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-13, IL-15, mouse IL-12 (p70) or mouse IL-12 (p40).

These human IL-12 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of both active (p70) and inactive (p40) forms of interleukin-12 (IL-12 total) in samples at 50 µL per well. Also included are calibrated recombinant protein standard, HRP-conjugate, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method: Colorimetric
Product Size: 96 assays
Protein Family: Cytokines & Receptors
Research Category: Immunology
Species: Human

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols