IL-6 Human ELISA Kit

Catalog number: EH2IL62

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Thermo Scientific Human IL-6 ELISA Kit is used to measure IL-6 (human interleukin-6) levels in human serum, plasma (EDTA, heparin, sodium citrate), urine and culture supernatants. Each kit contains two 96-well Strip Plates precoated with a human IL-6 capture antibody and sufficient detection antibody, conjugate, buffers, diluent, TMB and stop buffers, as well as lyophilized human IL-6 standard to analyze several samples.

Features of the Thermo Scientific Human IL-6 ELISA Kit:

Target: natural and recombinant human interleukin-6 (IL-6)
Format: colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: absorbance at 450nm minus absorbance at 550nm
Assay Range: 10.2 to 400 pg/mL
Sensitivity: < 1 pg/mL
Precision: intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: cell culture supernatant, urine, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 3 hours
Sample Size: 50 µL
Specificity: no significant cross-reaction with with human IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-7, IL-8, IL-10, IL-12, TNFα, IFNγ, IFNα, GM-CSF, or mouse IL-6.

Note: Assay range and other values listed above are for the pre-coated plate kit; see product documentation for values specific to the reagent kit and antibody-pair kit.

These human IL-6 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-6 (IL-6) in samples at 50 to 100 µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kits contain precoated 96-well strip plate(s) and lot-optimized reagents. The economically-priced reagent kit provides validated reagents only, requiring users to coat their own plates. The antibody-pair kit consists of the matched IL-6 antibody pairs and IL-6 standard only. All components undergo quality control testing to minimize lot-to-lot variability.

Includes:
Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a "Screening Set") contains matched antibodies, standard and detection reagents but no coated plates. The Antibody Pair Kit (previously called a "MiniKit") contains only matched antibodies and standard (no plates or detection reagents).

» Compare ELISA Kit formats »
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Sensitivity: See product documentation
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Pierce
Gene Aliases: BSF-2, BSF2, CDF, HGF, HSF, IFN-beta-2, IFNB2, IL-6
Gene ID: 3569
Gene Name: interleukin 6
Gene Symbol: IL6
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 2 x 96 tests
Protein Name: Interleukin-6
Target (Kit Named): IL-6
Target Species (Named): Human
Target Species (Validated): Human
UniProt ID: P05231

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols