What sample types have been validated for the Human Transglutaminase 2/TGM2 ELISA Kit (Cat. No. EH462RB)?
The Human Transglutaminase 2/TGM2 ELISA Kit (Cat. No. EH462RB) has been validated with serum, plasma, and supernatant sample types.