Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Human Angiogenin (Hu ANG) ELISA quantitates Hu ANG in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ANG.
Principle of the method
The Human ANG solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Angiogenin (ANG or ANG I) is important for the process of neovascularization and formation of new blood vessels. ANG is similar to pancreatic ribonuclease A and functions as a tRNA-specific ribonuclease that abolishes protein synthesis by specifically hydrolyzing cellular tRNAs. It interacts with endothelial cell-surface actin and may cause changes in the cell cytoskeleton. ANG is thought to be involved in the development of solid tumors and its antagonists are capable of inhibiting tumor growth. Defects in ANG are the cause of susceptibility to amyotrophic lateral sclerosis type 9 (ALS9). Angiogenin is a genetic link between ALS and PD.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Gene aliases : ALS9, ANG, HEL168, RAA1, RNASE4, RNASE5
Gene ID : (Human) 283
Gene symbol : ANG
Protein Aliases : Angiogenin, angiogenin, ribonuclease, RNase A family, 5, epididymis luminal protein 168, Ribonuclease 5, RNase 5, ribonuclease A A1, ribonuclease A family member 5
UniProt ID (Human) P03950
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support