Human FcGR2BC (CD32 B/C) ELISA Kit - FAQs
Human FcGR2BC (CD32 B/C) ELISA Kit - FAQs
View additional product information for Human FcGR2BC (CD32 B/C) ELISA Kit - FAQs (EHFCGR2BX10, EHFCGR2BX5, EHFCGR2B)