XCell6™ MultiGel Unit - FAQs
XCell6™ MultiGel Unit - FAQs
View additional product information for XCell6™ MultiGel Unit - FAQs (EI0006)
1 product FAQs found
Do you offer a unit for simultaneous running of up to 6 mini gels?
We used to offer the XCell6 MultiGel unit for simultaneous running of up to 6 mini gels, but it has been discontinued.
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.