Buffer Core with Cathode Bar for XCell SureLock™ Mini-Cell
Invitrogen™

Buffer Core with Cathode Bar for XCell SureLock™ Mini-Cell

Achieve fast, consistent protein gel separations in a small footprint with the updated Invitrogen XCell SureLock System. Its redesigned Buffer Core with a stainless-steel cathode bar increases run speed and system durability without compromising performance
Have Questions?
Catalog NumberQuantity
EI90151 Ea.
Catalog number EI9015
Price (EUR)
432,00
Each
Quantity:
1 Ea.
Price (EUR)
432,00
Each

The Invitrogen XCell SureLock Buffer Core is a key component of the XCell SureLock Mini-Cell vertical protein gel electrophoresis system. It provides a reliable electrical interface between the upper and lower buffer chambers, ensuring consistent and reproducible gel runs.

The updated Buffer Core design incorporates a stainless-steel cathode bar, platinum wire anode electrode, gold terminals, and durable silicone gaskets, offering a precise and leak-free seal during operation. This version features enhanced robustness and improved electrical efficiency for faster gel separations.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use WithProtein Gel Electrophoresis, 1D Gel Electrophoresis, SDS PAGE, Native PAGE, IEF
Gel TypeMini
Quantity1 Ea.
For Use With (Equipment)SureLock Mini
Product LineXCell
TypeElectrophoresis Core
Unit SizeEach

Frequently asked questions (FAQs)

Can I purchase replacement parts for the XCell SureLock Mini-Cell?

Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.