Product Image
Invitrogen™

Platinum Wire for the XCell SureLock™ apparatus (12 inch x 2)

Replacement Parts for Electrophoresis Apparatus자세히 알아보기
Have Questions?
카탈로그 번호수량
EI90221 Each
카탈로그 번호 EI9022
제품 가격(KRW)
140,000
온라인 행사
Ends: 30-Sep-2026
160,000
할인액 20,000 (13%)
Each
수량:
1 Each
제품 가격(KRW)
140,000
온라인 행사
Ends: 30-Sep-2026
160,000
할인액 20,000 (13%)
Each
Replacement Parts for Electrophoresis Apparatus
For Research Use Only. Not for use in diagnostic procedures.
사양
수량1 Each
배송 조건Room Temperature
용도(애플리케이션)1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
용도 (장비)XCell SureLock™ Mini
젤 크기Mini
제품라인XCell SureLock
유형Platinum Wire
Unit SizeEach
구성 및 보관
Store at room temperature.

자주 묻는 질문(FAQ)

Can I purchase replacement parts for the XCell SureLock Mini-Cell?

Here are the replacement parts we offer for the XCell SureLock Mini-Cell:
Replacement part - Cat. No.
Buffer Dam for XCell SureLock Mini-Cell and XCell II Blot Module - EI0012
L-Buffer Chamber for XCell SureLock Mini-Cell and XCell II Mini-Cell - EI0013
Platinum Wire for XCell SureLock Mini-Cell (12 inch x 2) - EI9022
XCell SureLock Lid - EI0010
XCell SureLock Wedge - EI0011
Buffer Core with Cathode Bar for XCell SureLock Mini-Cell - EI9015
Gel Knife - EI9010
Sponge Pad for Blotting (8) - EI9052

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

What are the components of the XCell SureLock Mini-Cell?

The XCell SureLock Mini-Cell includes the lid, gel tension wedge, lower buffer chamber, buffer core, buffer dam, and gel knife.

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.