Urea Nitrogen (BUN) Colorimetric Detection Kit
Invitrogen™

Urea Nitrogen (BUN) Colorimetric Detection Kit

The Urea Nitrogen (BUN) Detection research-use-only kit is a colorimetric assay designed for the quantification and detection of urea nitrogen詳細を見る
製品番号(カタログ番号)数量
EIABUN2 x 96 Tests
製品番号(カタログ番号) EIABUN
価格(JPY)
63,700
Each
数量:
2 x 96 Tests
The Urea Nitrogen (BUN) Detection research-use-only kit is a colorimetric assay designed for the quantification and detection of urea nitrogen in serum, plasma, urine, saliva and tissue culture media samples.

This complete, ready-to-use kit includes clear 96-well plate(s), urea nitrogen standard, and other components to perform the assay. A 96-well microplate reader capable of reading optical absorption at 450 nm is required for use of this kit.

Performance characteristics
• Assay type: colorimetric detection kit
• Sample types: serum, plasma, urine, saliva, and tissue culture media samples
• Sensitivity: 0.030 mg/dL
• Standard curve range: 0.156–10 mg/dL
• Reactivity: species independent

Background
Urea is a by-product of protein metabolism by the liver, and is therefore removed from the blood by the kidneys. Urea freely filters through the glomerulous, but is reabsorbed by the renal tubules in a flow-dependent fashion. The higher the flow rate, the greater amount of urea nitrogen is cleared from circulation and eliminated through the kidneys. Urea nitrogen is identical across all species and this kit will measure urea nitrogen from sources other than human.

Assay principle
The Urea Nitrogen ((also called BUN) Colorimetric Detection Kit is designed to quantitatively measure urea nitrogen in a variety of samples. A urea nitrogen standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Samples are mixed with color reagents A and B and incubated at room temperature for 30 minutes. The colored product is read at 450 nm.

Related links
Learn more about ELISA kits
Learn more about other immunoassays

研究用途にのみご使用ください。診断目的には使用できません。
仕様
使用対象 (装置)マイクロプレートリーダー
遺伝子ID(Entrez)非遺伝子
遺伝子記号非遺伝子
数量2 x 96 Tests
出荷条件氷水またはドライアイスでの出荷が承認されています
指名標的キット尿素窒素(BUN)
イムノアッセイキットのフォーマット検出キット
Unit SizeEach
組成および保存条件
96ウェルプレート、標準、その他のアッセイ特異的試薬、バッファー、ロット固有のテクニカルデータシート。

2–8℃で保管します。

よくあるご質問(FAQ)

Is the Urea Nitrogen (BUN) Colorimetric Detection Kit (Cat. No. EIABUN) based on an antibody-antigen reaction or chemical reaction?

The Urea Nitrogen (BUN) Colorimetric Detection Kit (Cat. No. EIABUN) is based on a chemical reaction, in which urea reacts with Reagent A to form a Schiff’s base, which then reacts with Reagent B to form a colored product.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.