Formaldehyde Fluorescent Detection Kit
Invitrogen™

Formaldehyde Fluorescent Detection Kit

The Formaldehyde Detection research-use-only kit is a fluorescent assay designed for the quantification and detection of formaldehyde in human urine자세히 알아보기
Have Questions?
카탈로그 번호수량
EIACH2O192 Tests
카탈로그 번호 EIACH2O
제품 가격(KRW)
635,000
온라인 행사
Ends: 30-Sep-2026
747,000
할인액 112,000 (15%)
Each
수량:
192 Tests
제품 가격(KRW)
635,000
온라인 행사
Ends: 30-Sep-2026
747,000
할인액 112,000 (15%)
Each
The Formaldehyde Detection research-use-only kit is a fluorescent assay designed for the quantification and detection of formaldehyde in human urine and tissue culture media samples.

This complete, ready-to-use kit includes clear 96-well plate(s), formaldehyde standard, formaldehyde detection reagent, and other components to perform the assay. A 96-well fluorescence plate reader capable of reading fluorescent emission at 510 nm, with excitation at 450 nm, is required for use of this kit.

Performance characteristics
• Assay type: fluorescent detection Kit
• Sample types: human urine and tissue culture media samples
• Sensitivity: 0.715 μM
• Standard curve range: 3.1–200 μM
• Reactivity: species independent

Background
Formaldehyde is widely employed in industry for a wide range of applications. It acts as a disinfectant and biocide, and is utilized as a tissue fixative and embalming agent. Formaldehyde is a key metabolite produced during normal metabolism and contributes to methylation reactions and protein and nucleic acid synthesis. This kit was shown to measure formaldehyde in human urine and tissue culture media. Formaldehyde is identical across all species and cell types.

Assay principle
The Formaldehyde Urinary Detection kit designed to quantitatively measure formaldehyde present in tissue culture media and urine samples. A formaldehyde standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a black microtiter plate. The fluorescent reaction is initiated with the formaldehyde detection reagent, which is pipetted into each well. After a short incubation the emission of the generated fluorescent signal is detected in a microtiter plate reader capable of measuring 510 nm fluorescence utilizing 450 nm excitation wavelength.

Related links
Learn more about ELISA kits
Learn more about other immunoassays

For Research Use Only. Not for use in diagnostic procedures.
사양
용도(장비)Microplate Reader
유전자 ID(Entrez)Non-Gene
유전자 기호Non-Gene
제품 유형Formaldehyde Fluorescent Detection Kit
수량192 Tests
배송 조건Approved for shipment on Wet or Dry Ice
타겟 키트 이름 지정Formaldehyde
이뮤노어세이 키트 형식Fluorescent Detection Kit
샘플 종류Urine
Unit SizeEach
구성 및 보관
96-well plate(s), standard, other assay specific reagents, buffers, and lot-specific technical data sheet.

Store at –20°C.