Search
View additional product information for Thyroxine (T4) Competitive ELISA Kit - FAQs (EIAT4CX10, EIAT4C)
1 product FAQs found
Both these kits recognized their intended targets (thyroxine and triiodothyronine) independent of species. The reason this assay works for all species is because these small hormones have highly conserved sequences among species.