Heratherm Incubation Chambers
Heratherm Incubation Chambers
Heratherm Incubation Chambers
Heratherm Incubation Chambers
Heratherm Incubation Chambers
Heratherm Incubation Chambers
Heratherm Incubation Chambers
Thermo Scientific™

Heratherm Incubation Chambers

Comprehensive data monitoring and temperature control combined with a flexible Heratherm Incubation Chamber provide a dependable option for microbiological incubation or temperature-dependent storage and testing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDoor StyleCapacity (Metric)
EIS750Solid (outer), Glass (inner)750 L
EIG750Glass750 L
EIS250Solid outer door with inner glass door247 L
EIG250Glass door247 L
Catalog number EIS750
Price (BRL)
-
Door Style:
Solid (outer), Glass (inner)
Capacity (Metric):
750 L
Produces precise temperature conditions above and below ambient, ideally suited for microbiological incubation, and heated or cooled incubated storage.
  • Uniform Temperature throughout the chamber.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • Intuitive touchscreen interface with easy programmability of temperature cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, Ethernet connection, or Thermo Cloud via OPC-UA standard.
  • Sample and process protection: Assign personalized access with user IDs; restrict access with standard door lock, or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Glass door also available.
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
  • Duplex interior electrical sockets for use of stirrers, shakers or other equipment inside the chamber are available as a factory-installed option. Choose from a variety of country-specific sockets. (Ordered with separate order code)
Specifications
Alarm TypesAudible and Visible
Capacity (English)26.5 cu.ft.
Capacity (English) Shelf66.1 lbs
Capacity (Metric) Shelf30 kg
Certifications/ComplianceUL, CE
Data OutputsUSB, Ethernet
Depth (English)34.84 in.
Depth (English) Interior24.8 in.
Depth (Metric)885 mm
Depth (Metric) Interior630 mm
Display7 in. touchscreen user interface
Door StyleSolid (outer), Glass (inner)
Electrical Requirements100/240V, 50/60 Hz, 1400W
Height (English)72.24 in.
Height (English) Interior57.09 in
Height (Metric)1835 mm
Height (Metric) Interior1450 mm
LightingNo
No. of Casters4
Plug TypeNema 5-20P
Ports2 access ports
ProgrammabilityTemperature ramping
Resolution0.1°C
Shelf Area (English)4.9 ft.²
Shelf Area (Metric)0.46 m²
Shelf MaterialStainless Steel (304/1.4301)
Shipping Weight (English)840 lbs
Shipping Weight (Metric)381 kg
Temperature ControlMicroprocessor - programmable
Temperature Stability≤ ±0.1°C @ 20°C to 37°C
Temperature Uniformity≤ ±0.5°C @ 20°C to 37°C
TypeEnvironmental Chamber
Warranty24 months
Wattage900 W
Width (English)42.52 in.
Width (English) Interior33.07 in.
Width (English) Shelf22.6 in.
Width (Metric)1080 mm
Width (Metric) Interior840 mm
Width (Metric) Shelf795 mm
Capacity (Metric)750 L
Chamber MaterialPowder-coated Cold Rolled Steel
MaterialPowder-coated Cold Rolled Steel
No. of Shelves3
Temperature Range (Metric)0 to 70°C
Voltage100/115/220/230 V
Unit SizeEach