Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Heratherm Light Chambers 750 L
Thermo Scientific™

Heratherm Light Chambers 750 L

Accurate and even light distribution with Long-lasting LED lighting for (ICH) light testing, plant growth and animal hatching — relative humidity and temperature are controlled to meet specified environment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDoor StyleFor Use With (Application)Includes
ELG750Glass doorPlant growth, light testing, animal hatchingLight modules not included - for plant growth/testing/animal hatching: 333980G01
ELS750Solid outer door with inner glass doorICH light testing, plant growth, light testing, animal hatchingLight modules not included - ICH testing: 333979G01 / Standard: 333980G01
Catalog number ELG750
Price (MXN)
-
Door Style:
Glass door
For Use With (Application):
Plant growth, light testing, animal hatching
Includes:
Light modules not included - for plant growth/testing/animal hatching: 333980G01
Accurate control of three parameters - Temperature above and below ambient, relative humidity and light - provide great environment in testing applications and for biological research with plants and animals. Choice of ICH module provides capability for efficient ICH light testing in Pharma.
  • Uniform Temperature / relative humidity / light exposure for samples.
  • Energy-efficient Peltier technology supports temperature stability and uniformity with lower energy usage compared to traditional compressors and without hazardous refrigerant.
  • LED light system provides specified light exposure on 1, 2 or 3 shelves; programmable to adapt light quality / intensity and on/off timing - e.g. for day-night cycles. Choose from two light modules (ordered separately):
    • 1. ICH module, includes UV LED for ICH light testing;can only be used in ELS750 - model with solid outer door - for safety reasons
    • 2. Testing module for plant growth / daylight testing / animal hatching
  • Environmentally friendly water-efficient humidification system - with water tank or optional fix water connection for in-house purified water
  • Intuitive touchscreen interface with easy programmability of parameter cycles and process duration.
  • Two access ports for independent data capture, and internal documentation of set and actual parameter values, along with event log to meet 21 CFR Part 11 requirements for process documentation.
  • Data retrieval via USB stick, or Thermo Cloud via OPC-UA standard (with Connect Edge solution)
  • Sample and process protection: Assign personalized access with passcode; restrict access with standard key door lock or optional automated door lock; various alarm options to prevent operator of process issues.
  • Simplified setup with no technician required; install using standard power cord.
  • IQ/OQ qualifications and other services available; visit thermofisher.com/chamberservice.
  • Inner glass door keeps environment stable, even when opening outer door. Exterior glass door also available (not for ICH module use - for safety reasons).
  • Rounded corners and removable shelving offer easy cleaning to help prevent cross-contamination.
Specifications
Alarm TypesAudible and Visible
Capacity (English)26.5 cu.ft
Capacity (English) Shelf66 lbs (stationary)
Capacity (Metric) Shelf30 kg (stationary)
Certifications/ComplianceUL, CE
Data OutputsUSB, Thermo Cloud via OPC UA
Dimensions (L x W x H)895 x 1080 x 1835 mm / 35.24 x 42.52 x 72.24 in
Dimensions (LxWxH) Interior630 x 840 x 1450 mm / 24.8 x 33.07 x 57.09 in
Display7 in. Touchscreen user interface
Door StyleGlass door
Electrical Requirements100 - 240V, 50-60Hz, 1700W
For Use With (Application)Plant growth, light testing, animal hatching
Humidity Range10 - 80% RH with light; 10 -90% RH w/o light
Humidity Resolution0.1% RH
Humidity SourceRemovable water tank
IncludesLight modules not included - for plant growth/testing/animal hatching: 333980G01
Length (Metric) Shelf795 mm
No. of Casters4
Plug TypeNema 5-20P
Ports2 access ports
ProgrammabilityTemperature, relative humidity, light intensity
Shelf Area (English)4.9 ft2

Shelf Area (Metric)0.46 m2
Shelf MaterialStainless Steel (304/1.4301)
Shipping Weight (English)842 lbs
Shipping Weight (Metric)382 kg
Temperature ControlMicroprocessor - programmable
Temperature Resolution0.1°C
Temperature Stability≤ ±0.3°C @ 25°C to 40°C
Temperature Uniformity≤ ±2.0°C @ 25°C to 40°C
TypeVertical Light Chamber
Warranty24 months
Water Consumption0.2- 1.5 L per day; Value depends on settings, usage (e.g. number of door openings), and environmental conditions
Water Resistivity0.05 - 1 MΩ
Wattage1700 W
Width (English) Shelf31.3 in.
Capacity (Metric)750 L
Chamber MaterialStainless Steel (304/1.4301)
MaterialPowder-coated Galvanized Steel
No. of Shelves3
Relative Humidity10 to 90%
Temperature Range (Metric)15 to 45°C with light; 5 to 60°C without light
Voltage100/240 V
Unit SizeEach