Elite PCTS pH / Conductivity / TDS / Salinity Pocket Testers and Replacement Sensors
Elite PCTS pH / Conductivity / TDS / Salinity Pocket Testers and Replacement Sensors
Thermo Scientific™

Elite PCTS pH / Conductivity / TDS / Salinity Pocket Testers and Replacement Sensors

Flexible on-the-go pH, conductivity, TDS, salinity and temperature testing using durable Elite PCTS pocket testers for a variety of applications including agriculture, aquaculture, hydroponics and more!
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호제품 유형
ELITEPCTSKITPCTS Tester Kit
01X001530Replacement PCTS Sensor
ELITEPCTSPCTS Tester
카탈로그 번호 ELITEPCTSKIT
제품 가격(KRW)
-
제품 유형:
PCTS Tester Kit
Benefit from these pocket-sized testers. Thermo Scientific™ Elite testers are designed to provide on-the-go measurements for a variety of applications including homebrew beer and wine, food and beverage, hydroponics, aquaculture and aquariums, agriculture, pools and spas, environmental monitoring, education and more. The Elite CTS cup-sensor pocket testers are ideal for flexible on-the-go conductivity, TDS, salinity and temperature testing in a compact, travel-ready design.

Elite Pocket Testers offer flexible on-the-go electrochemical testing. Choose the Elite PCTS model to measure pH, conductivity, total dissolved solids (TDS) or salinity with temperature for a wide range of applications. Elite pocket testers are durable, with a large backlit screen, one-touch calibration feature and replaceable sensor heads. They are ideal for testing beverages including homebrew beer and wine, food, hydroponics, aquaculture and aquariums, environmental monitoring and more.

  • Flexible testing option
  • Measure pH, conductivity, TDS or salinity with temperature
  • Large back-lit graphic display
  • Uses 4 x AAA alkaline batteries
  • Average 150 hours of battery life
  • Wide leak-proof cap serves as stable base
  • Replaceable sensors
  • One-year warranty for tester
  • Six-month warranty for replacement sensors
사양
정확도(전도도)± 1% full scale
정확도(염도)± 1% full scale
정확도(TDS)± 1% full scale
정확도(pH)±0.02
백라이트 옵션Yes
배터리 수명150 hr. (average)
배터리 종류4 x AAA Alkaline
보정 모드pH, conductivity, TDS, salinity, temperature
보정 지점(전도도)1 to 3
보정 지점(pH)1 to 3
인증/적합성CE, FCC
설명Elite PCTS pocket tester for measuring pH, conductivity, TDS or salinity with temperature
포함Elite PCTS tester with batteries, lanyard, soft carry case, pH buffers and conductivity standard
제품 유형PCTS Tester Kit
상대 습도 범위5 to 85 %, non-condensing
분해능(전도도)0.1 μS/cm for 0.0 to 200.0 μS/cm; 1 μS/cm for200 to 2000 μS/cm; 0.01 mS/cm for 2.00 to 20.00 mS/cm
분해능(염도)0.10 ppt
분해능(TDS)0.1 ppm for 0.0 to 100.0 ppm; 1 ppm for 100 to 1000 ppm; 0.01 ppt for 0.10 to 10.00 ppt
염도 측정 모드Per practical salinity curve
안정성0.40 to 1.00 (selectable)
TDS 계수 범위0.40 to 1.00 (selectable)
온도 정확도±0.6°C, ±1.0°F for 0.0 to 50.0°C; ±1.1°C, ±1.9°F for 50.1 to 60.0°C
온도 보상Yes
작동 온도(미터법) 작동5°C to 45°C (ambient)
온도 프로브 보정1 point
온도 범위(영국식 단위)32.0°F to 140.0°F
온도 분해능0.1°C, 0.1°F
워런티1 Year for Tester
디스플레이 유형Graphic Dot Matrix
IP 등급IP67
Measurement ModespH, Conductivity, TDS, Salinity, Temperature
범위(전도도)0.00 to 20.00 mS/cm
범위(염도)0.00 to 10.00 ppt
범위(TDS)0.00 to 10.00 ppt
범위(pH)-1.00 to 15.00
분해능(pH)0.01
온도 범위(미터법)0.0°C to 60.0°C
Unit SizeEach