Elite pH Spear Pocket Testers and Replacement Sensors
Thermo Scientific™
Elite pH Spear Pocket Testers and Replacement Sensors
Flexible on-the-go pH testing using durable Elite spear pocket testers with replaceable sensors for a variety of applications include food, beverage, agriculture and more!
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호제품 유형
ELITEPHSPEARKITpH Spear Pocket Tester Kit
01X001538Replacement Sensor
ELITEPHSPEARpH Spear Pocket Tester
카탈로그 번호 ELITEPHSPEARKIT
제품 가격(KRW)
-
제품 유형:
pH Spear Pocket Tester Kit
Benefit from these pocket-sized testers. Thermo Scientific™ Elite testers are designed to provide on-the-go measurements for a variety of applications including homebrew beer and wine, food and beverage, hydroponics, aquaculture and aquariums, agriculture, pools and spas, environmental monitoring, education and more. The Elite CTS cup-sensor pocket testers are ideal for flexible on-the-go conductivity, TDS, salinity and temperature testing in a compact, travel-ready design.
Elite Pocket Testers offer flexible on-the-go electrochemical testing. Choose the Elite pH Spear model to measure pH using a spear-shaped sensor design for your application. Elite pocket testers are durable, with a large backlit screen, one-touch calibration feature and replaceable sensor heads. They are ideal for testing beverages including homebrew beer and wine, food, hydroponics, aquaculture and aquariums, environmental monitoring and more.
  • Flexible testing option
  • Measure pH
  • Large back-lit graphic display
  • Uses 4 x AAA alkaline batteries
  • Average 250 hours of battery life
  • Wide leak-proof cap serves as stable base
  • Replaceable sensors
  • One-year warranty for tester
  • Six-month warranty for replacement sensors
사양
정확도(pH)±0.02
백라이트 옵션Yes
배터리 수명250 hr. (average)
배터리 종류4 x AAA Alkaline
보정pH
보정 지점1 to 3
인증/적합성CE, FCC
채널1
설명Elite pH spear pocket tester for measuring pH with a spear-tip sensor
포함Elite pH spear tester with batteries, lanyard, and pH buffer
제품 유형pH Spear Pocket Tester Kit
상대 습도 범위5 to 85 %, non-condensing
분해능(pH)0.01
안정성Stability icon on display
온도 정확도Manual input
온도 보상Yes with manual temperature input
작동 온도(미터법) 작동5°C to 45°C (ambient)
온도 범위(영국식 단위)32.0°F to 140.0°F
온도 분해능0.1°C, 0.1°F
워런티1 Year for Tester
디스플레이 유형Graphic Dot Matrix
IP 등급IP67
Measurement ModespH
범위(염도)0 to 80 ppt
범위(pH)-1 to 15
온도 범위(미터법)0.0°C to 60.0°C
Unit SizeEach