IL-6 Mouse ELISA Kit

Catalog number: EM2IL65

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Assay IL-6 (mouse interleukin-6) levels in mouse serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific Mouse IL-6 ELISA Kit.

Features of the Mouse IL-6 ELISA Kit:

Target: natural and recombinant mouse interleukin-6 (IL-6)
Format: colorimetric ELISA (TMB substrate) in clear 96-well strip plates
Measurement: absorbance at 450nm minus absorbance at 550nm
Assay Range: 51 to 2000 pg/mL
Sensitivity: < 7 pg/mL
Precision: intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 4 hours
Sample Size: 50 µL
Specificity: no significant cross-reaction with mouse GM-CSF, IFNγ, IL-1α, IL-2, IL-3, IL-4, IL-5, IL-7, IL-9, IL-10, IL-12, or human IL-6. This kit has been shown to have a high level of cross-reactivity with recombinant rat IL-6.

This mouse IL-6 ELISA kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-6 (IL-6) in samples at 50 to 100 µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kit contains pre-coated 96-well strip plates and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

» Compare ELISA Kit formats »
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Sensitivity: See product documentation
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Pierce
Gene Aliases: IL-6
Gene ID: 16193
Gene Name: interleukin 6
Gene Symbol: IL6
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 5 x 96 tests
Protein Aliases: IL-6, B-cell hybridoma growth factor, Interleukin HP-1
Protein Name: Interleukin-6
Target (Kit Named): IL-6
Target Species (Named): Mouse
Target Species (Validated): Mouse
UniProt ID: P08505

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols