GM-CSF Mouse ELISA Kit

Catalog number: EMGMCSF5

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Assay GM-CSF (granulocyte macrophage colony-stimulating factor) levels in mouse serum and culture supernatants with our Thermo Scientific Mouse GM-CSF ELISA Kit.

Features of the Thermo Scientific Mouse GM-CSF ELISA Kit:

Target: natural and recombinant mouse granulocyte-macrophage colony-stimulating factor (GM-CSF
Format: colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: absorbance at 450nm minus absorbance at 550nm
Assay Range: 0 to 250 pg/mL
Sensitivity: < 5 pg/mL
Reproducibility: intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Sample Types: cell culture supernatant, serum
Total Assay Time: 3.5 hours
Sample Size: 50 µL
Specificity: no significant cross-reaction with mouse IL-2, IL-3, IL-4, IL-5, IL-6, IFNγ, TNFα, rabbit GM-CSF, rat GM-CSF or human GM-CSF.

This mouse GM-CSF ELISA kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of granulocyte macrophage colony-stimulating factor (GM-CSF) in samples at 50 µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kit contains pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

Includes:
Complete kit contains pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Sensitivity: See product documentation
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Pierce
Gene Aliases: Csfgm, GMCSF, Gm-CSf, MGI-IGM
Gene ID: 12981
Gene Name: colony stimulating factor 2 (granulocyte-macrophage)
Gene Symbol: CSF2
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 5 x 96 tests
Protein Aliases: GM-CSF, Colony-stimulating factor, CSF
Protein Name: Granulocyte-macrophage colony-stimulating factor
Target (Kit Named): GM-CSF
Target Species (Named): Mouse
Target Species (Validated): Mouse
UniProt ID: P01587

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols