Mouse IL-12 p40/70 (IL-12B) ELISA Kit - FAQs
Mouse IL-12 p40/70 (IL-12B) ELISA Kit - FAQs
View additional product information for Mouse IL-12 p40/70 (IL-12B) ELISA Kit - FAQs (EMIL12BX10, EMIL12B, EMIL12BX5)