Mouse TIMP1-Cell Lysate ELISA Kit - FAQs
Mouse TIMP1-Cell Lysate ELISA Kit - FAQs
View additional product information for Mouse TIMP1-Cell Lysate ELISA Kit - FAQs (EMTIMP1CL, EMTIMP1CLX10, EMTIMP1CLX5)