TNF alpha Mouse ELISA Kit

Catalog number: EMTNFA5

Invitrogen™  Related applications: Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Assay TNFα (mouse tumor necrosis factor alpha) levels in mouse serum and culture supernatant samples with our Thermo Scientific Mouse TNF alpha ELISA Kit.

Features of the Mouse TNF alpha ELISA Kit:

Target: natural and recombinant mouse TNFα (mouse tumor necrosis factor alpha)
Format: colorimetric ELISA (TMB substrate) in clear 96-well strip plates
Measurement: absorbance at 450nm minus absorbance at 550nm
Assay Range: 50 to 2450 pg/mL
Sensitivity: < 9 pg/mL
Precision: intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: cell culture supernatant, serum
Total Assay Time: 3 hours
Sample Size: 50 µL
Specificity: no significant cross-reaction with mouse IL-2, IL-3, IL-4, IL-5, IL-6, GM-CSF, IFNγ or human TNFα. This ELISA has some cross-reactivity with rabbit and cynomologous monkey TNFα in culture supernatants.

This mouse TNF alpha ELISA kit uses pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of tumor necrosis factor alpha (TNF-alpha, TNFα) in samples at 50 to 100 µL per well. Also included are calibrated recombinant protein standard, streptavidin-HRP, TMB substrate solution and stop solution. The complete kit contains precoated 96-well strip plates and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.

» Compare ELISA Kit formats »

TNFα, also called cachectin, macrophage cytotoxin, necrosin, etc., is produced primarily by macrophages in response to bacteria, viruses, mycoplasma, immune complexes, other cytokines, tumor cells, complement and phorbol esters among others. It is a 157 amino acid long polypeptide with an apparent MW of 17 kDa under denaturing conditions but exists as a 45-55 kDa trimer naturally.

In vivo, TNFα has anti-tumor activities against some solid tumors, induces tumorigenesis, metastisis; acts in immunomodulation; induces septic shock, fever; plays a role in multiple sclerosis; and is involved in autoimmunity and infectious disease. TNFα alone and in combination with other cytokines and chemical agents has been used as an anti-tumor agent. Patients with melanoma and other solid tumors, administered high doses of TNFα, have realized a cure rate of up to 90% albeit with some major and minor side effects.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Sensitivity: See product documentation
Detection Method: Colorimetric
For Use With (Equipment): Microplate Reader
Former Identifiers: Pierce
Gene Aliases: DIF, TNF-a, TNF-alpha, TNFSF2, TNFalphaa, Tnfsf1a, Tnlg1f, Tnf
Gene ID: 21926
Gene Name: tumor necrosis factor
Gene Symbol: TNF
Immunoassay Kit Format: Coated ELISA Kit
Label or Dye: HRP (Horseradish Peroxidase)
Product Size: 5 x 96 tests
Protein Name: tumor necrosis factor
Target (Kit Named): TNF alpha
Target Species (Named): Mouse
Target Species (Validated): Mouse
UniProt ID: P06804

Contents & storage

Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.

Documents

Manuals & protocols