Mouse BAFF R (TNFRSF13C) ELISA Kit - FAQs
Mouse BAFF R (TNFRSF13C) ELISA Kit - FAQs
View additional product information for Mouse BAFF R (TNFRSF13C) ELISA Kit - FAQs (EMTNFRSF13CX5, EMTNFRSF13CX10, EMTNFRSF13C)