Uracil-DNA Glycosylase (1 U/μL)
Thermo Scientific™

Uracil-DNA Glycosylase (1 U/μL)

Thermo Scientific Uracil-DNA Glycosylase (UDG, UNG) catalyzes the hydrolysis of the N-glycosylic bond between uracil and sugar, leaving an apyrimidinicRead more
Catalog NumberQuantity
EN0361200 units
EN03625 x 200 units
Catalog number EN0361
Price (JPY)
10,700
Online offer
Ends: 25-Dec-2026
17,900
Save 7,200 (40%)
Each
Quantity:
200 units
Request bulk or custom format
Thermo Scientific Uracil-DNA Glycosylase (UDG, UNG) catalyzes the hydrolysis of the N-glycosylic bond between uracil and sugar, leaving an apyrimidinic site in uracil-containing single or double-stranded DNA (see Figure 1 in Supporting Data). The enzyme shows no activity on RNA.

Highlights

• Active in Thermo Scientific buffers for restriction enzymes and thermophilic polymerases

Applications

• Control of carry-over contamination in PCR
• Glycosylase mediated single nucleotide polymorphism detection (GMPD)
• Site-directed mutagenesis
• As a probe for protein-DNA interaction studies
• SNP genotyping
• Cloning of PCR products
• Generation of single strand overhangs of PCR products and cDNA

Note

The abasic sites formed in DNA by Uracil-DNA Glycosylase may be subsequently cleaved by heat, alkali-treatment or endonucleases that cleave specifically at abasic sites. UDG (UNG) is active in the presence or absence of divalent cations.

Use of this enzyme in certain applications may be covered by patents and may require a license.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Buffer10X Reaction Buffer
Product TypeUracil DNA Glycosylase
Quantity200 units
Concentration1 U/μL
EnzymeUracil-DNA Glycosylase
Unit SizeEach
Contents & Storage
Uracil-DNA glycolase (1 U/μL) and 10X Reaction Buffer. Store at -20°C.

Frequently asked questions (FAQs)

What is UDG?

UDG (uracil-DNA-glycosylase) refers to a superfamily of enzymes comprising six sub-families. Family I UDG enzymes are called UNG, after the uracil-N-glycosylase gene. The terms UDG and UNG are commonly used interchangeably because they perform the same function in qPCR, namely, to remove Uracil from dU-containing DNA to prevent carryover contamination from previous PCRs. See the following link: https://www.thermofisher.com/uk/en/home/life-science/pcr/real-time-pcr/real-time-pcr-learning-center/real-time-pcr-basics/what-is-ung-udg.html