DNase I, RNase-free (1 U/μL) - FAQs
View additional product information for DNase I, RNase-free (1 U/μL) - FAQs (EN0521)
1 product FAQs found
Does your DNase I, RNase-free (Cat. No. EN0521) have any tags?
This product has no tags.
Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.