RNase A, DNase and protease-free (10 mg/mL) - FAQs
RNase A, DNase and protease-free (10 mg/mL) - FAQs
View additional product information for RNase A, DNase and protease-free (10 mg/mL) - FAQs (EN0531)
1 product FAQs found
What is the molecular weight of your RNAse A, DNAse and Protease-free endoribonuclease (Cat. No. EN0531)?
The molecular weight is 13.5 kDa.