RiboLock RNase Inhibitor (40 U/μL), 24 x 2,500 units - FAQs
View additional product information for RiboLock RNase Inhibitor (40 U/μL) - FAQs (EO0384, EO0381, EO0382)
3 product FAQs found
What are the recommended reaction time and temperature for inactivating RNAse A with Thermo Scientific RiboLock RNase Inhibitor?
The “activity” of RiboLock RNAse Inhibitor in principle is temperature-independent because there are no chemical reactions involved. At both 25 degrees C and 37 degrees C, the same quantity of RiboLock RNAse Inhibitor is needed to quench a given amount of RNase A.
What is the molecular weight of Thermo Scientific RiboLock RNase Inhibitor?
The molecular weight of the monomer is 49.6 kDa.
Which RNases does Thermo Scientific RiboLock RNase Inhibitor inhibit?
Thermo Scientific RiboLock RNase Inhibitor inhibits the activity of RNases A, B, and C. It does not inhibit eukaryotic RNases T1, T2, U1, U2, CL3 or prokaryotic RNases I and H.