Search
Buscar
View additional product information for Proteinase K, recombinant, PCR grade - FAQs (EO0491, EO0492)
1 product FAQs found
The recommended working concentration of Proteinase K is 0.05 to 1 mg/mL.