Proteinase K, recombinant, PCR grade, 5 x 1.0 mL - FAQs
Proteinase K, recombinant, PCR grade, 5 x 1.0 mL - FAQs
View additional product information for Proteinase K, recombinant, PCR grade - FAQs (EO0491, EO0492)
1 product FAQs found
What is the recommended working concentration of Proteinase K reagent?
The recommended working concentration of Proteinase K is 0.05 to 1 mg/mL.