T4 DNA Polymerase (5 U/μL)
Thermo Scientific™

T4 DNA Polymerase (5 U/μL)

Thermo Scientific T4 DNA Polymerase is a template-dependent DNA polymerase that catalyzes 5'-3' synthesis from primed single-stranded DNA. The enzymeRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EP0062500 Units
EP0061100 Units
Catalog number EP0062
Price (USD)
268.65
Online exclusive
295.00
Save 26.35
Each
Quantity:
500 Units
Request bulk or custom format
Price (USD)
268.65
Online exclusive
295.00
Save 26.35
Each

Thermo Scientific T4 DNA Polymerase is a template-dependent DNA polymerase that catalyzes 5'-3' synthesis from primed single-stranded DNA. The enzyme has a 3'-5' exonuclease activity, but lacks 5'-3' exonuclease activity.

Highlights

  • Stronger 3'-5' exonuclease activity on single-stranded than on double-stranded DNA and greater (more than 200 times) than DNA polymerase I (Cat. No. EP0041), E. coli,and Klenow fragment (Cat. No. EP0051)
  • Active in Thermo Scientific restriction enzyme, PCR, RT and T4 DNA Ligase (Cat. No. EL0011) buffers

Applications

  • Blunting of DNA ends: fill-in of 5'-overhangs or/and removal of 3'-overhangs(see References1, 2)
  • Blunting of PCR products with 3'-dA overhangs
  • Synthesis of labeled DNA probes by the replacement reaction(see Reference3)
  • Oligonucleotide-directed site-specific mutagenesis(see Reference4)
  • Ligation-independent cloning of PCR products
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Includes2 x 1 mL of 5X Reaction Buffer
PolymeraseT4 DNA Polymerase
Quantity500 Units
Concentration5 U/μL
Unit SizeEach