Taq DNA Polymerase, native (5 U/μL)
Thermo Scientific™

Taq DNA Polymerase, native (5 U/μL)

Thermo Scientific Native Taq DNA Polymerase is a highly thermostable DNA polymerase of the thermophilic bacterium Thermus aquaticus.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormat
EP0072With BSA
EP0282Without BSA
Catalog number EP0072
Price (MXN)
-
Format:
With BSA
Request bulk or custom format
Thermo Scientific Taq DNA Polymerase, native, is a highly thermostable DNA polymerase of the thermophilic bacterium Thermus aquaticus. The enzyme catalyzes 5' to 3' synthesis of DNA, has no detectable 3' to 5' exonuclease (proofreading) activity, and possesses low 5' to 3' exonuclease activity. In addition, the polymerase exhibits deoxynucleotidyl transferase activity, which frequently results in the addition of extra adenines at the 3'-end of PCR products. Native Taq DNA Polymerase is preferred for amplification of bacterial DNA sequences homologous to those found in E. coli.

Available with or without the stabilizing agent BSA, Taq DNA Polymerase, native, with BSA, is often considered the best choice when amplifying DNA samples of lower purity, e.g., genomic DNA from mouse tail.

Both native Taq DNA polymerases are supplied with two buffers: 10X Taq Buffer with KCl and 10X Taq Buffer with (NH4)2SO4. The latter allows for PCR at a wide range of magnesium concentrations and decreases unspecific priming

Features

  • Thermostable with half-life more than 40 min at 95°C
  • Generates PCR products with 3'-dA overhangs
  • Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescent-labeled nucleotides)

Applications

  • Routine PCR amplification of DNA fragments up to 5 kb
  • DNA labeling
  • High throughput PCR

Notes

  • The error rate of Taq DNA Polymerase in PCR is 2.2 x 10-5 errors per nt per cycle. Accordingly, the accuracy of PCR is 4.5 x 104. Accuracy is an inverse of the error rate and shows an average number of correct nucleotides incorporated before an error occurs.
  • The 10X Taq Buffer without Detergent is recommended for microarray experiments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatWith BSA
Optimal Reaction Temperature72°C
PolymeraseTaq DNA Polymerase
Product TypeDNA Polymerase
Quantity500 units
Research CategoryPCR
Shipping ConditionDry Ice
Concentration5 U/μL
For Use With (Application)Standard PCR
Unit SizeEach
Contents & Storage
• Taq DNA Polymerase, native (5 U/μL), 100 μL
• 10X Taq Buffer with KCl, 2 x 1.25 mL
• 10X Taq Buffer with (NH4)2SO4, 2 x 1.25 mL
• 25 mM MgCl2, 2 x 1.25 mL
• BSA (20 mg/mL), 0.25 mL

Store at -20°C.