RNA Polymerases
Thermo Scientific™

RNA Polymerases

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPolymeraseConcentrationQuantity
EP0111AFT7 RNA Polymerase200 U/μL20,000 U
EP168AFPoly(A) RNA Polymerase5 U/μL800 U
Catalog number EP0111AF
Price (USD)
304.80
Special offer
Online exclusive
Ends: 27-Jun-2026
381.00
Save 76.20 (20%)
Each
Polymerase:
T7 RNA Polymerase
Concentration:
200 U/μL
Quantity:
20,000 U
Price (USD)
304.80
Special offer
Online exclusive
Ends: 27-Jun-2026
381.00
Save 76.20 (20%)
Each

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration200 U/μL
Quantity20,000 U
PolymeraseT7 RNA Polymerase
Product LineTheraPure™ GMP
PromoterT7
Unit SizeEach
Contents & Storage
200 U/μL T7 RNA Polymerase, 100 μL

Store at -20°C.