Upgrade to Maxima™ H Minus Reverse Transcriptase for better performance. List price reduced by 40%.
RevertAid Reverse Transcriptase (200 U/μL)
Thermo Scientific™

RevertAid Reverse Transcriptase (200 U/μL)

Thermo Scientific RevertAid Reverse Transcriptase (RT) is a recombinant M-MuLV RT. It differs from the M-MuLV RT by its structureRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
EP044110,000 Units
EP04425 x 10,000 Units
Catalog number EP0441
Price (USD)
105.65
Online exclusive
126.00
Save 20.35
Each
Quantity:
10,000 Units
Request bulk or custom format
Price (USD)
105.65
Online exclusive
126.00
Save 20.35
Each

Thermo Scientific RevertAid Reverse Transcriptase (RT) is a recombinant M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity and a RNase H activity specific to RNA in RNA-DNA hybrids that is significantly lower than that of Avian Myeloblastosis Virus (AMV) reverse transcriptase.

Features of RevertAid Reverse Transcriptase include:
• Efficient synthesis of full-length first strand cDNA up to 13 kb
• Optimum activity at 42°C
• Active up to 50°C
• Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, and fluorescein-labeled nucleotides)

Applications
• First strand cDNA synthesis for RT-PCR and real-time RT-PCR
• Synthesis of cDNA for cloning and expression
• Generation of labeled cDNA probes for microarrays
• DNA labeling
• Analysis of RNA by primer extension

Source
RevertAid Reverse Transcriptase is sourced from E.coli cells with a cloned fragment of the pol gene encoding Moloney Murine Leukemia Virus reverse transcriptase.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatStand-alone Enzyme
Optimal Reaction Temperature42°C
Quantity10,000 Units
Reaction FormatSeparate components
Reagent TypeReverse Transcription
Reverse TranscriptaseRevertAid
Ribonuclease H ActivityYes
Shipping ConditionDry Ice
Size (Final Product)Up to 13 kb
Starting MaterialRNA
TechniqueReverse Transcription
Concentration200 U/μL
Reaction Speed60 min.
Unit SizeEach
Contents & Storage

• RevertAid Reverse Transcriptase (200 U/μL)
• 5X Reaction Buffer

Store at –20°C.

Frequently asked questions (FAQs)

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H-minus RT or Maxima H-minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H-minus RTs for template-independent addition of C nucleotides.

What steps should I take while performing first strand cDNA synthesis using low purity template (e.g., inhibitors in RNA sample)?

Trace amounts of reagents used in RNA purification protocols may remain in solution and inhibit first-strand synthesis, e.g., SDS, EDTA, guanidine salts, phosphate, pyrophosphate, polyamines, spermidine. To remove trace contaminants, we recommend re-precipitating the RNA with ethanol and washing the pellet with 75% ethanol, or re-purifying the RNA.

For reverse transcription, how important is the quality of RNA template?

RNA purity and integrity are essential for synthesis and quantification of cDNA. Always assess the integrity of RNA prior to cDNA synthesis. Use freshly prepared RNA. Multiple freeze/thaw cycles of the RNA sample and synthesized cDNA is not recommended. Avoid RNase contamination and discard low quality RNA.

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H Minus RT or Maxima H Minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H Minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in 1-step RT-qPCR applications.

Do Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess terminal deoxynucleotidyl (TdT) activity?

All Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess intrinsic TdT activity, although at varying degrees depending upon the reaction conditions. We recommend specialized SuperScript IV Template Switching RT Master Mix for high efficiency in applications requiring template switching RT.