Maxima Hot Start Taq DNA Polymerase (5 U/µL)

Catalog number: EP0602

Thermo Scientific™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Maxima Hot Start Taq DNA Polymerase is a chemically modified recombinant Taq DNA polymerase. The enzyme is inactive at room temperature, avoiding extension of non-specifically annealed primers or primer dimers and providing higher specificity of DNA amplification. The functional activity of the enzyme is restored during a short 4 minute incubation at 95°C. The activated enzyme maintains the same functionality as Taq DNA polymerase. Maxima Hot Start Taq DNA Polymerase is also available in a master mix format.

Highlights


• Four minute activation time
• High PCR specificity and sensitivity
• Room temperature PCR set up

Applications

• Hot Start PCR
• Routine PCR
• High throughput PCR
• Multiplex PCR
• Genotyping
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: Maxima Hot Start Taq DNA Polymerase
Hot Start: Built-In Hot Start
Fidelity (vs. Taq): 1 X
GC-Rich PCR Performance: Low
Reaction Format: Standalone
Product Overhang: 3'-A
Reaction Speed: Standard
Product Line: Maxima
Product Size: 500 units

Contents & storage

Contains:
• 100 µL Maxima Hot Start Taq DNA Polymerase (5 U/µL)
• 2 x 1.25 mL 10X Hot Start PCR Buffer
• 2 x 1.25 mL 25 mM MgCl2

Store at -20°C.

Documents

Manuals & protocols

Recommended products