Bsm DNA Polymerase, large fragment (8 U/μL)
Thermo Scientific™

Bsm DNA Polymerase, large fragment (8 U/μL)

Thermo Scientific Bsm DNA Polymerase, Large Fragment, is equivalent to Bst DNA polymerase, which catalyzes 5' → 3' synthesis ofRead more
Have Questions?
Catalog NumberQuantity
EP06911,600 units
Catalog number EP0691
Price (USD)
84.65
Online exclusive
118.00
Save 33.35
Each
Quantity:
1,600 units
Request bulk or custom format
Price (USD)
84.65
Online exclusive
118.00
Save 33.35
Each

Thermo Scientific Bsm DNA Polymerase, Large Fragment, is equivalent to Bst DNA polymerase, which catalyzes 5' → 3' synthesis of DNA and lacks 5' → 3' and 3' → 5' exonuclease activities. Sourced from E. coli cells with a cloned polA gene from Bacillus smithii, Bsm DNA Polymerase, Large Fragment, features strong strand displacement activity and is active in a wide range of temperatures (30–63°C), with an optimum of activity at 60°C. With high functional similarity to Bst DNA Polymerase, Large Fragment, this enzyme can serve as an alternative in most applications, including loop-mediated isothermal amplification (LAMP).

Applications
Bsm DNA Polymerase, Large Fragment, may be used in a wide variety of applications including:
• Loop-mediated isothermal amplification (LAMP)
• Whole genome amplification (WGA)
• Ramification amplification (RAM)
• Random-primed DNA labeling
• Labeling by fill-in 5'-overhangs of dsDNA

Discover the power of loop-mediated isothermal amplification ›

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration8 U/μL
Optimal Reaction Temperature60°C
Reaction Time30 to 60 minutes
Shipping ConditionDry Ice
For Use With (Application)LAMP, RT-LAMP, WGA, RAM
PolymeraseBsm DNA Polymerase
Product TypeStand-alone enzyme
Quantity1,600 units
Unit SizeEach
Contents & Storage

• Bsm DNA Polymerase, Large Fragment (200 μL at 8 U/μL)
• 10X Bsm Buffer (1.25 mL)

Store at –15 to –25°C.