DreamTaq DNA Polymerase (5 U/μL), 10 x 500 Units - FAQs
View additional product information for DreamTaq DNA Polymerase (5 U/μL) - FAQs (EP0702, EP0713, EP0703, EP0714, EP0704, EP0705, EP0711, EP0701, EP0712)
1 product FAQs found
What is the error rate of DreamTaq DNA Polymerases?
The error rate of DreamTaq DNA Polymerase is the same as that of Taq DNA Polymerase: 2.2x10^-5 errors per nt per cycle.